missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Iodo-L-phenylalanine, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5197903
Description
4-Iodo-L-phenylalanine may be used in protein engineering as a model unnatural α amino acid to alter primary amino acid composition via the opal (UGA) codon.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Iodo-L-phenylalanine may be used in protein engineering as a model unnatural α amino acid to alter primary amino acid composition via the opal (UGA) codon.
Solubility
Partly miscible with water.
Notes
Light sensitive. Incompatible with oxidizing agents.
Specifications
4-Iodo-L-phenylalanine | |
C9H10INO2 | |
4411317 | |
Partly miscible with water. | |
N[C@@H](CC1=CC=C(I)C=C1)C(O)=O | |
291.09 | |
291.09 | |
1 g | |
Light sensitive |
24250-85-9 | |
MFCD00002602 | |
4-iodo-l-phenylalanine, 4-iodophenylalanine, l-4-iodophenylalanine, iodo-phenylalanine, p-iodo-l-phenylalanine, p-iodophenylalanine, s-2-amino-3-4-iodophenyl propanoic acid, 2s-2-amino-3-4-iodophenyl propanoic acid, h-phe 4-i-oh, phenylalanine, 4-iodo | |
PZNQZSRPDOEBMS-QMMMGPOBSA-N | |
(2S)-2-amino-3-(4-iodophenyl)propanoic acid | |
134497 | |
95% | |
−6.5° (c=1 in Acetic Acid:Water (4:1)) |
Chemical Identifiers
24250-85-9 | |
291.09 | |
PZNQZSRPDOEBMS-QMMMGPOBSA-N | |
134497 | |
N[C@@H](CC1=CC=C(I)C=C1)C(O)=O |
C9H10INO2 | |
MFCD00002602 | |
4-iodo-l-phenylalanine, 4-iodophenylalanine, l-4-iodophenylalanine, iodo-phenylalanine, p-iodo-l-phenylalanine, p-iodophenylalanine, s-2-amino-3-4-iodophenyl propanoic acid, 2s-2-amino-3-4-iodophenyl propanoic acid, h-phe 4-i-oh, phenylalanine, 4-iodo | |
(2S)-2-amino-3-(4-iodophenyl)propanoic acid |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only