Learn More
4-Iodo-o-phenylenediamine, 95%, Thermo Scientific Chemicals
$213.66 - $843.70
Chemical Identifiers
CAS | 21304-38-1 |
---|---|
Molecular Formula | C6H7IN2 |
Molecular Weight (g/mol) | 234.04 |
MDL Number | MFCD08669466 |
InChI Key | FUOSRKZBOIVBOS-UHFFFAOYSA-N |
Synonym | 1,2-diamino-4-iodobenzene, 4-iodo-1,2-diaminobenzene, 4-iodo-1,2-phenylenediamine, 1,2-benzenediamine, 4-iodo, 4-iodo-1,2-benzenediamine, 4-iodo-benzene-1,2-diamine, ambkkkkk335, 2-amino-5-iodoaniline, 2-amino4-iodophenylamine |
PubChem CID | 11207049 |
IUPAC Name | 4-iodobenzene-1,2-diamine |
SMILES | NC1=C(N)C=C(I)C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6446903
|
Thermo Scientific Chemicals
H6446903 |
1 g |
Each for $213.66
|
|
|||||
AAH6446906
|
Thermo Scientific Chemicals
H6446906 |
5 g |
Each for $843.70
|
|
|||||
Description
It is employed in the synthesis of N-(Benzimidazol-2-ylmethyl)iminodiacetic acids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed in the synthesis of N-(Benzimidazol-2-ylmethyl)iminodiacetic acids.
Solubility
Slightly soluble in water.
Notes
Light sensitive. Store away from light, acids and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
21304-38-1 | |
234.04 | |
FUOSRKZBOIVBOS-UHFFFAOYSA-N | |
11207049 | |
NC1=C(N)C=C(I)C=C1 |
C6H7IN2 | |
MFCD08669466 | |
1,2-diamino-4-iodobenzene, 4-iodo-1,2-diaminobenzene, 4-iodo-1,2-phenylenediamine, 1,2-benzenediamine, 4-iodo, 4-iodo-1,2-benzenediamine, 4-iodo-benzene-1,2-diamine, ambkkkkk335, 2-amino-5-iodoaniline, 2-amino4-iodophenylamine | |
4-iodobenzene-1,2-diamine |
Specifications
21304-38-1 | |
C6H7IN2 | |
1 g | |
2716284 | |
1,2-diamino-4-iodobenzene, 4-iodo-1,2-diaminobenzene, 4-iodo-1,2-phenylenediamine, 1,2-benzenediamine, 4-iodo, 4-iodo-1,2-benzenediamine, 4-iodo-benzene-1,2-diamine, ambkkkkk335, 2-amino-5-iodoaniline, 2-amino4-iodophenylamine | |
FUOSRKZBOIVBOS-UHFFFAOYSA-N | |
4-iodobenzene-1,2-diamine | |
11207049 | |
95% | |
4-Iodo-o-phenylenediamine |
73°C to 77°C | |
MFCD08669466 | |
UN2811 | |
Light sensitive | |
Slightly soluble in water. | |
NC1=C(N)C=C(I)C=C1 | |
234.04 | |
234.04 | |
Solid |
Safety and Handling
GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only