missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Iodobenzoic acid, 98%, Thermo Scientific™

Chemical Identifiers

CAS 619-58-9
Molecular Formula C7H5IO2
Molecular Weight (g/mol) 248.02
MDL Number MFCD00002533
InChI Key GHICCUXQJBDNRN-UHFFFAOYSA-N
Synonym p-iodobenzoic acid, benzoic acid, 4-iodo, benzoic acid, p-iodo, 4-iodo-benzoic acid, p-iodobenzenecarboxylic acid, 4-jodbenzoesaeure, unii-ipo4lyq1en, 4-jodbenzoesaeure german, 4-iodo benzoic acid, ipo4lyq1en
PubChem CID 12085
IUPAC Name 4-iodobenzoic acid
SMILES OC(=O)C1=CC=C(I)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC124480100
SDS View Documents
Thermo Scientific Chemicals
124480100
10g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC124480500
SDS View Documents
Thermo Scientific Chemicals
124480500
50g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

619-58-9
248.02
GHICCUXQJBDNRN-UHFFFAOYSA-N
12085
OC(=O)C1=CC=C(I)C=C1
C7H5IO2
MFCD00002533
p-iodobenzoic acid, benzoic acid, 4-iodo, benzoic acid, p-iodo, 4-iodo-benzoic acid, p-iodobenzenecarboxylic acid, 4-jodbenzoesaeure, unii-ipo4lyq1en, 4-jodbenzoesaeure german, 4-iodo benzoic acid, ipo4lyq1en
4-iodobenzoic acid

Specifications

619-58-9
100.0
Authentic
Glass bottle
IC6H4COOH
10g
Solubility in water: 0.04g/L (25°C)
OC(=O)C1=CC=C(I)C=C1
248.02
248.02
4-Iodobenzoic acid, 98%
97.5
>110°C
97.5% min. (HPLC)
C7H5IO2
MFCD00002533
p-iodobenzoic acid, benzoic acid, 4-iodo, benzoic acid, p-iodo, 4-iodo-benzoic acid, p-iodobenzenecarboxylic acid, 4-jodbenzoesaeure, unii-ipo4lyq1en, 4-jodbenzoesaeure german, 4-iodo benzoic acid, ipo4lyq1en
GHICCUXQJBDNRN-UHFFFAOYSA-N
4-iodobenzoic acid
12085
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 210-603-2

RTECSNumber : DH2976000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only