missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Iodobenzonitrile, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2510414

 View more versions of this product

Catalog No. AAB2510414


Explore available promotions

Description

Description

4-Iodobenzonitrile is used in organic synthesis. It is used as catalytic agent, petrochemical additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodobenzonitrile is used in organic synthesis. It is used as catalytic agent, petrochemical additive.

Solubility
Insoluble in water.

Notes
Light Sensitive. Store in the dark. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
Specifications

Specifications

4-Iodobenzonitrile
125°C to 127°C
MFCD00051310
UN3439
p-iodobenzonitrile, benzonitrile, 4-iodo, p-cyanoiodobenzene, benzonitrile, p-iodo, 1-cyano-4-iodobenzene, 4-cyanoiodobenzene, 4-iodo-benzonitrile, 4-iodobenzenecarbonitrile, 4-iodbenzonitril, 4-iodo benzonitrile
XOKDXPVXJWTSRM-UHFFFAOYSA-N
4-iodobenzonitrile
76467
98%
3058-39-7
C7H4IN
25 g
Light sensitive
Insoluble in water.
IC1=CC=C(C=C1)C#N
229.02
229.02

Chemical Identifiers

3058-39-7
229.02
XOKDXPVXJWTSRM-UHFFFAOYSA-N
76467
IC1=CC=C(C=C1)C#N
C7H4IN
MFCD00051310
p-iodobenzonitrile, benzonitrile, 4-iodo, p-cyanoiodobenzene, benzonitrile, p-iodo, 1-cyano-4-iodobenzene, 4-cyanoiodobenzene, 4-iodo-benzonitrile, 4-iodobenzenecarbonitrile, 4-iodbenzonitril, 4-iodo benzonitrile
4-iodobenzonitrile
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H312-H332-H315-H319-H335
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only