Learn More
4-Iodobenzylamine hydrochloride, 97%, Thermo Scientific Chemicals
$141.16 - $476.63
Chemical Identifiers
CAS | 59528-27-7 |
---|---|
Molecular Formula | C7H9ClIN |
Molecular Weight (g/mol) | 269.51 |
MDL Number | MFCD04039324 |
InChI Key | GBJMURRFWZREHE-UHFFFAOYSA-N |
Synonym | 4-iodobenzylamine hydrochloride, 4-iodophenyl methanamine hydrochloride, 4-iodobenzylamine hcl, benzenemethanamine, 4-iodo-, hydrochloride, 4-iodophenyl methylamine, chloride, 1-4-iodophenyl methanamine hydrochloride, 4-iodobenzylamine, hydrochloride salt, 4-iodobenzylaminehydrochloride, pubchem3304, acmc-209tzk |
PubChem CID | 16217505 |
IUPAC Name | (4-iodophenyl)methanamine;hydrochloride |
SMILES | C1=CC(=CC=C1CN)I.Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAL1996703
|
Thermo Scientific Chemicals
L1996703 |
1 g |
N/A
|
|
||||||
AAL1996706
|
Thermo Scientific Chemicals
L1996706 |
5 g |
N/A
|
|
||||||
Description
4-Iodobenzylamine hydrochloride is used to prepare cyclic amide by reacting with methyl 4-bromomethyl-3-methoxycarbonylcinnamate in the presence of triethylamine. It is also used in pharmaceuticals, agrochemicals and dye industries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Iodobenzylamine hydrochloride is used to prepare cyclic amide by reacting with methyl 4-bromomethyl-3-methoxycarbonylcinnamate in the presence of triethylamine. It is also used in pharmaceuticals, agrochemicals and dye industries.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
59528-27-7 | |
269.51 | |
GBJMURRFWZREHE-UHFFFAOYSA-N | |
16217505 | |
C1=CC(=CC=C1CN)I.Cl |
C7H9ClIN | |
MFCD04039324 | |
4-iodobenzylamine hydrochloride, 4-iodophenyl methanamine hydrochloride, 4-iodobenzylamine hcl, benzenemethanamine, 4-iodo-, hydrochloride, 4-iodophenyl methylamine, chloride, 1-4-iodophenyl methanamine hydrochloride, 4-iodobenzylamine, hydrochloride salt, 4-iodobenzylaminehydrochloride, pubchem3304, acmc-209tzk | |
(4-iodophenyl)methanamine;hydrochloride |
Specifications
59528-27-7 | |
C7H9ClIN | |
1 g | |
Light sensitive | |
Soluble in water. | |
C1=CC(=CC=C1CN)I.Cl | |
269.51 | |
269.51 | |
4-Iodobenzylamine hydrochloride |
275°C to 280°C (decomposition) | |
MFCD04039324 | |
3691299 | |
4-iodobenzylamine hydrochloride, 4-iodophenyl methanamine hydrochloride, 4-iodobenzylamine hcl, benzenemethanamine, 4-iodo-, hydrochloride, 4-iodophenyl methylamine, chloride, 1-4-iodophenyl methanamine hydrochloride, 4-iodobenzylamine, hydrochloride salt, 4-iodobenzylaminehydrochloride, pubchem3304, acmc-209tzk | |
GBJMURRFWZREHE-UHFFFAOYSA-N | |
(4-iodophenyl)methanamine;hydrochloride | |
16217505 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only