Learn More
4-Iodophenol, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1345536
Description
4-Iodophenol is a chemical used in chemiluminescence imaging assays and experiments, as diagnostic agents in order to detect cancer cells in patients. Also it is used in the synthesis of agonists towards the estrogen β receptor. Also used in wide range of medicals industrial applications as well as in human and animal nutrition products such as pharmaceutical intermediates, and polarizing films for Liquid Crystal Display (LCD) chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Iodophenol is a chemical used in chemiluminescence imaging assays and experiments, as diagnostic agents in order to detect cancer cells in patients. Also it is used in the synthesis of agonists towards the estrogen β receptor. Also used in wide range of medicals industrial applications as well as in human and animal nutrition products such as antiseptics and disinfectants, pharmaceutical intermediates, polarizing films for liquid crystal display (LCD) chemicals.
Solubility
Soluble in alcohol and ether. Slightly soluble in water.
Notes
Light Sensitive, store in dark. Store in cool, dry place in tightly closed container. Store away from acid chlorides and acid anhydrides.
Specifications
4-Iodophenol | |
91°C to 95°C | |
C6H5IO | |
500 g | |
Light sensitive | |
p-iodophenol, phenol, 4-iodo, phenol, p-iodo, 4-hydroxyiodobenzene, p-hydroxyiodobenzene, 4-hydroxyphenyl iodide, 4-iodo-phenol, unii-bh194bak0b, ccris 668, bh194bak0b | |
VSMDINRNYYEDRN-UHFFFAOYSA-N | |
4-iodophenol | |
10894 | |
220.01 |
540-38-5 | |
138°C (5 mmHg) | |
MFCD00002327 | |
1904544 | |
14,5036 | |
Soluble in alcohol and ether. Slightly soluble in water. | |
OC1=CC=C(I)C=C1 | |
220.01 | |
CHEBI:43521 | |
≥98% |
Chemical Identifiers
540-38-5 | |
220.01 | |
VSMDINRNYYEDRN-UHFFFAOYSA-N | |
10894 | |
4-iodophenol |
C6H5IO | |
MFCD00002327 | |
p-iodophenol, phenol, 4-iodo, phenol, p-iodo, 4-hydroxyiodobenzene, p-hydroxyiodobenzene, 4-hydroxyphenyl iodide, 4-iodo-phenol, unii-bh194bak0b, ccris 668, bh194bak0b | |
CHEBI:43521 | |
OC1=CC=C(I)C=C1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312+H332-H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: III
EINECSNumber : 208-745-5
RTECSNumber : SL5600000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only