missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Iodophenylboronic acid, 97%, Thermo Scientific Chemicals

$63.34 - $162.02

Chemical Identifiers

CAS 5122-99-6
Molecular Formula C6H6BIO2
Molecular Weight (g/mol) 247.83
MDL Number MFCD01319014
InChI Key PELJYVULHLKXFF-UHFFFAOYSA-N
Synonym 4-iodophenyl boronic acid, 4-iodobenzeneboronic acid, p-iodophenylboronic acid, 4-iodophenylboronicacid, p-iodobenzeneboronic acid, p-iodophenyl boronic acid, boronic acid, 4-iodophenyl, 4-boronoiodobenzene, pubchem6160, pubchem11574
PubChem CID 151254
IUPAC Name (4-iodophenyl)boronic acid
SMILES OB(O)C1=CC=C(I)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC359480010
SDS View Documents
Thermo Scientific Chemicals
359480010
1 g Glass bottle N/A
 
AC359480050
SDS View Documents
Thermo Scientific Chemicals
359480050
5 g Glass bottle N/A
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5122-99-6
247.83
PELJYVULHLKXFF-UHFFFAOYSA-N
151254
OB(O)C1=CC=C(I)C=C1
C6H6BIO2
MFCD01319014
4-iodophenyl boronic acid, 4-iodobenzeneboronic acid, p-iodophenylboronic acid, 4-iodophenylboronicacid, p-iodobenzeneboronic acid, p-iodophenyl boronic acid, boronic acid, 4-iodophenyl, 4-boronoiodobenzene, pubchem6160, pubchem11574
(4-iodophenyl)boronic acid

Specifications

5122-99-6
100.0
Beige
96% min. (HPLC)
C6H6BIO2
1 g
Solubility in water: 25g/L
OB(O)C1=CC=C(I)C=C1
247.83
247.82
Powder
96.0
322.0°C to 326.0°C
Authentic
Glass bottle
MFCD01319014
4-iodophenyl boronic acid, 4-iodobenzeneboronic acid, p-iodophenylboronic acid, 4-iodophenylboronicacid, p-iodobenzeneboronic acid, p-iodophenyl boronic acid, boronic acid, 4-iodophenyl, 4-boronoiodobenzene, pubchem6160, pubchem11574
PELJYVULHLKXFF-UHFFFAOYSA-N
(4-iodophenyl)boronic acid
151254
97%
4-Iodophenylboronic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only