Learn More
4-Isobutyl-alpha-methylphenylacetic acid, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2098914
Description
Ibuprofen is useful as a nonsteroidal anti-inflammatory research chemical. Ibuprofen is reported to inhibit the netrophil-dependent edema response of rabbit skin to C5a/PGE23. NSAID that inhibits cyclooxygenase-1 and -2.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIbuprofen is useful as a nonsteroidal anti-inflammatory research chemical. Ibuprofen is reported to inhibit the netrophil-dependent edema response of rabbit skin to C5a/PGE23. NSAID that inhibits cyclooxygenase-1 and -2.
Solubility
Insoluble in water. Soluble in ethanol (25 mg/ml), chloroform (1:1), ether (1:2), acetone (1:1.5), aqueous solutions of alkali hydroxides and carbonates, dichloromethane, methanol (50 mg/ml), and ethyl acetate.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
4-Isobutyl-alpha-methylphenylacetic acid | |
75°C to 78°C | |
Odorless | |
MFCD00010393 | |
UN3077 | |
ibuprofen, 2-4-isobutylphenyl propanoic acid, motrin, brufen, nurofen, advil, dolgit, liptan, nuprin, anflagen | |
HEFNNWSXXWATRW-UHFFFAOYNA-N | |
2-[4-(2-methylpropyl)phenyl]propanoic acid | |
3672 | |
206.29 |
15687-27-1 | |
157°C (4 mmHg) | |
C13H18O2 | |
25 g | |
14,4881 | |
Insoluble in water. Soluble in ethanol (25mg/ml),chloroform (1:1),ether (1:2),acetone (1:1.5),aqueous solutions of alkali hydroxides and carbonates,dichloromethane,methanol (50mg/ml),and ethyl acetate. | |
CC(C)CC1=CC=C(C=C1)C(C)C(O)=O | |
206.29 | |
CHEBI:5855 | |
99% |
Chemical Identifiers
15687-27-1 | |
206.29 | |
HEFNNWSXXWATRW-UHFFFAOYNA-N | |
3672 | |
2-[4-(2-methylpropyl)phenyl]propanoic acid |
C13H18O2 | |
MFCD00010393 | |
ibuprofen, 2-4-isobutylphenyl propanoic acid, motrin, brufen, nurofen, advil, dolgit, liptan, nuprin, anflagen | |
CHEBI:5855 | |
CC(C)CC1=CC=C(C=C1)C(C)C(O)=O |
Safety and Handling
GHS H Statement
H301-H361
Toxic if swallowed.
Suspected of damaging fertility or the unborn child.
P260-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P314-P330-P501c
H302-H319-H335
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 239-784-6
RTECSNumber : MU6640000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only