Learn More
4-Isopropoxybenzenesulfonyl chloride, 96%, Thermo Scientific Chemicals
$62.84 - $157.42
Chemical Identifiers
CAS | 98995-40-5 |
---|---|
Molecular Formula | C9H11ClO3S |
Molecular Weight (g/mol) | 234.694 |
MDL Number | MFCD03093083 |
InChI Key | IJWCRHKAQNFJLT-UHFFFAOYSA-N |
Synonym | 4-isopropoxybenzenesulfonyl chloride, 4-isopropoxybenzene-1-sulfonyl chloride, 4-isopropoxyphenylsulfonyl chloride, 4-propan-2-yloxy benzene-1-sulfonyl chloride, benzenesulfonyl chloride, 4-1-methylethoxy, 4-isopropoxybenzenesulphonyl chloride, benzenesulfonyl chloride, 4-1-methylethoxy-9ci, 4-isopropoxybenzenesulfonylchloride, pubchem19078, acmc-209sbk |
PubChem CID | 22327297 |
IUPAC Name | 4-propan-2-yloxybenzenesulfonyl chloride |
SMILES | CC(C)OC1=CC=C(C=C1)S(=O)(=O)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1898403
|
Thermo Scientific Chemicals
L1898403 |
1 g |
Each for $62.84
|
|
|||||
AAL1898406
|
Thermo Scientific Chemicals
L1898406 |
5 g |
Each for $157.42
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
98995-40-5 | |
234.694 | |
IJWCRHKAQNFJLT-UHFFFAOYSA-N | |
22327297 | |
CC(C)OC1=CC=C(C=C1)S(=O)(=O)Cl |
C9H11ClO3S | |
MFCD03093083 | |
4-isopropoxybenzenesulfonyl chloride, 4-isopropoxybenzene-1-sulfonyl chloride, 4-isopropoxyphenylsulfonyl chloride, 4-propan-2-yloxy benzene-1-sulfonyl chloride, benzenesulfonyl chloride, 4-1-methylethoxy, 4-isopropoxybenzenesulphonyl chloride, benzenesulfonyl chloride, 4-1-methylethoxy-9ci, 4-isopropoxybenzenesulfonylchloride, pubchem19078, acmc-209sbk | |
4-propan-2-yloxybenzenesulfonyl chloride |
Specifications
98995-40-5 | |
112°C to 113°C (0.01 mmHg) | |
MFCD03093083 | |
UN3261 | |
Moisture sensitive | |
IJWCRHKAQNFJLT-UHFFFAOYSA-N | |
4-propan-2-yloxybenzenesulfonyl chloride | |
22327297 | |
96% |
30°C to 37°C | |
C9H11ClO3S | |
1 g | |
3269058 | |
4-isopropoxybenzenesulfonyl chloride, 4-isopropoxybenzene-1-sulfonyl chloride, 4-isopropoxyphenylsulfonyl chloride, 4-propan-2-yloxy benzene-1-sulfonyl chloride, benzenesulfonyl chloride, 4-1-methylethoxy, 4-isopropoxybenzenesulphonyl chloride, benzenesulfonyl chloride, 4-1-methylethoxy-9ci, 4-isopropoxybenzenesulfonylchloride, pubchem19078, acmc-209sbk | |
CC(C)OC1=CC=C(C=C1)S(=O)(=O)Cl | |
234.694 | |
234.7 | |
4-Isopropoxybenzenesulfonyl chloride, 25g |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only