missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Isopropylbenzaldehyde, 98%, Thermo Scientific Chemicals

$113.09 - $113.09

Chemical Identifiers

CAS 122-03-2
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.21
MDL Number MFCD00006953
InChI Key WTWBUQJHJGUZCY-UHFFFAOYSA-N
Synonym 4-isopropylbenzaldehyde, cuminaldehyde, cuminic aldehyde, cumaldehyde, cuminal, p-cumic aldehyde, cumic aldehyde, cuminyl aldehyde, p-isopropylbenzaldehyde, benzaldehyde, 4-1-methylethyl
PubChem CID 326
ChEBI CHEBI:28671
IUPAC Name 4-propan-2-ylbenzaldehyde
SMILES CC(C)C1=CC=C(C=O)C=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC334881000
SDS View Documents
Thermo Scientific Chemicals
334881000
100 g Glass bottle
Each for $113.09
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

122-03-2
148.21
WTWBUQJHJGUZCY-UHFFFAOYSA-N
326
4-propan-2-ylbenzaldehyde
C10H12O
MFCD00006953
4-isopropylbenzaldehyde, cuminaldehyde, cuminic aldehyde, cumaldehyde, cuminal, p-cumic aldehyde, cumic aldehyde, cuminyl aldehyde, p-isopropylbenzaldehyde, benzaldehyde, 4-1-methylethyl
CHEBI:28671
CC(C)C1=CC=C(C=O)C=C1

Specifications

122-03-2
100.0
0.9790g/mL
93°C
97.5% min. (GC)
C10H12O
MFCD00006953
0.979
4-isopropylbenzaldehyde, cuminaldehyde, cuminic aldehyde, cumaldehyde, cuminal, p-cumic aldehyde, cumic aldehyde, cuminyl aldehyde, p-isopropylbenzaldehyde, benzaldehyde, 4-1-methylethyl
WTWBUQJHJGUZCY-UHFFFAOYSA-N
4-propan-2-ylbenzaldehyde
326
148.2
Liquid
97.5
Colorless to Yellow
234°C to 235°C
Authentic
Glass bottle
1.5290 to 1.532
100 g
15, 2611
Solubility in water: insoluble
CC(C)C1=CC=C(C=O)C=C1
148.21
CHEBI:28671
98%
4-Isopropylbenzaldehyde, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Warning

EINECSNumber : 204-516-9

RTECSNumber : CU7000000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only