missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Isopropylbenzaldehyde, tech. 90%, Thermo Scientific Chemicals

$56.25 - $321.41

Chemical Identifiers

CAS 122-03-2
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.21
MDL Number MFCD00006953
InChI Key WTWBUQJHJGUZCY-UHFFFAOYSA-N
Synonym 4-isopropylbenzaldehyde, cuminaldehyde, cuminic aldehyde, cumaldehyde, cuminal, p-cumic aldehyde, cumic aldehyde, cuminyl aldehyde, p-isopropylbenzaldehyde, benzaldehyde, 4-1-methylethyl
PubChem CID 326
ChEBI CHEBI:28671
SMILES CC(C)C1=CC=C(C=O)C=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1401914
View Documents
Thermo Scientific Chemicals
A1401914
25 g
Each for $56.25
 
AAA1401922
View Documents
Thermo Scientific Chemicals
A1401922
100 g
Each for $105.18
 
AAA1401936
View Documents
Thermo Scientific Chemicals
A1401936
500 g
Each for $321.41
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

122-03-2
148.21
WTWBUQJHJGUZCY-UHFFFAOYSA-N
326
CC(C)C1=CC=C(C=O)C=C1
C10H12O
MFCD00006953
4-isopropylbenzaldehyde, cuminaldehyde, cuminic aldehyde, cumaldehyde, cuminal, p-cumic aldehyde, cumic aldehyde, cuminyl aldehyde, p-isopropylbenzaldehyde, benzaldehyde, 4-1-methylethyl
CHEBI:28671

Specifications

122-03-2
234°C to 235°C
C10H12O
MFCD00006953
636547
14,2621
WTWBUQJHJGUZCY-UHFFFAOYSA-N
4-(propan-2-yl)benzaldehyde
326
148.21
Technical
0.979
93°C (199°F)
1.53
25 g
Air Sensitive
4-isopropylbenzaldehyde, cuminaldehyde, cuminic aldehyde, cumaldehyde, cuminal, p-cumic aldehyde, cumic aldehyde, cuminyl aldehyde, p-isopropylbenzaldehyde, benzaldehyde, 4-1-methylethyl
CC(C)C1=CC=C(C=O)C=C1
148.21
CHEBI:28671
90%
4-Isopropylbenzaldehyde
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H227-H315-H319-H335
Toxic if swallowed.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P264b-P270-P280-P301+P312-P330-P370+P378q-P501c

H227-H302

EINECSNumber : 204-516-9

RTECSNumber : CU7000000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only