missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Isopropylbenzoic acid, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 412800250

 View more versions of this product

Catalog No. AC412800250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Isopropylbenzoic acid
98.0
116.0°C to 120.0°C
Authentic
Glass bottle
(CH3)2CHC6H4COOH
25 g
4-isopropylbenzoic acid, cuminic acid, cumic acid, p-isopropylbenzoic acid, 4-1-methylethyl benzoic acid, benzoic acid, 4-1-methylethyl, 4-propan-2-yl benzoic acid, benzoic acid, p-isopropyl, p-cumic acid, 4-isopropyl benzoic acid
CKMXAIVXVKGGFM-UHFFFAOYSA-N
4-propan-2-ylbenzoic acid
10820
164.2
Crystalline Powder
536-66-3
100.0
White
98% min. (HPLC)
C10H12O2
MFCD00002564
15, 2608
Solubility in water: sparingly soluble
CC(C)C1=CC=C(C=C1)C(=O)O
164.2
CHEBI:28122
98+%

Chemical Identifiers

536-66-3
164.2
CKMXAIVXVKGGFM-UHFFFAOYSA-N
10820
4-propan-2-ylbenzoic acid
C10H12O2
MFCD00002564
4-isopropylbenzoic acid, cuminic acid, cumic acid, p-isopropylbenzoic acid, 4-1-methylethyl benzoic acid, benzoic acid, 4-1-methylethyl, 4-propan-2-yl benzoic acid, benzoic acid, p-isopropyl, p-cumic acid, 4-isopropyl benzoic acid
CHEBI:28122
CC(C)C1=CC=C(C=C1)C(=O)O
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 208-642-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only