Learn More
4-Isopropylbenzoyl chloride, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H3664506
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Isopropylbenzoyl chloride | |
Yellow | |
MFCD03424703 | |
UN3265 | |
4-isopropylbenzoyl chloride, 4-1-methylethyl benzoyl chloride, 4-isopropyl-benzoyl chloride, 4-iso-propyl-benzoyl chloride, 4-1-methylethyl-benzoyl chloride, 4-propan-2-yl benzoyl chloride, benzoyl chloride, 4-1-methylethyl, p-isopropylbenzoyl chloride, rho-isopropylbenzoyl chloride, 4-isopropyl benzoyl chloride | |
CC(C)C1=CC=C(C=C1)C(=O)Cl | |
182.647 | |
182.65 | |
Liquid |
21900-62-9 | |
C10H11ClO | |
5 g | |
Moisture sensitive | |
LBSYWDTVBUZMCM-UHFFFAOYSA-N | |
4-propan-2-ylbenzoyl chloride | |
2759486 | |
97% |
Chemical Identifiers
21900-62-9 | |
182.647 | |
LBSYWDTVBUZMCM-UHFFFAOYSA-N | |
2759486 | |
CC(C)C1=CC=C(C=C1)C(=O)Cl |
C10H11ClO | |
MFCD03424703 | |
4-isopropylbenzoyl chloride, 4-1-methylethyl benzoyl chloride, 4-isopropyl-benzoyl chloride, 4-iso-propyl-benzoyl chloride, 4-1-methylethyl-benzoyl chloride, 4-propan-2-yl benzoyl chloride, benzoyl chloride, 4-1-methylethyl, p-isopropylbenzoyl chloride, rho-isopropylbenzoyl chloride, 4-isopropyl benzoyl chloride | |
4-propan-2-ylbenzoyl chloride |
Safety and Handling
GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312+H332-H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: II
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only