Learn More
4-Mercaptobenzoic acid, 90%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 373300050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Mercaptobenzoic acid | |
88.0 | |
223.0°C to 225.0°C | |
Glass bottle | |
HSC6H4CO2H | |
5 g | |
LMJXSOYPAOSIPZ-UHFFFAOYSA-N | |
4-sulfanylbenzoic acid | |
95738 | |
90% |
1074-36-8 | |
100.0 | |
88% min. (HPLC) | |
C7H6O2S | |
MFCD00016617 | |
4-mercaptobenzoic acid, 4-mercaptobenzoate, benzoic acid, 4-mercapto, 4-mercapto-benzoic acid, 4-mercapto benzoic acid, 4-thiobenzoic acid, p-mercaptobenzoic acid, benzoic acid, p-mercapto, para-mercaptobenzoate, paracarboxythiophenol | |
OC(=O)C1=CC=C(S)C=C1 | |
154.18 | |
154.19 |
Chemical Identifiers
1074-36-8 | |
154.18 | |
LMJXSOYPAOSIPZ-UHFFFAOYSA-N | |
95738 | |
OC(=O)C1=CC=C(S)C=C1 |
C7H6O2S | |
MFCD00016617 | |
4-mercaptobenzoic acid, 4-mercaptobenzoate, benzoic acid, 4-mercapto, 4-mercapto-benzoic acid, 4-mercapto benzoic acid, 4-thiobenzoic acid, p-mercaptobenzoic acid, benzoic acid, p-mercapto, para-mercaptobenzoate, paracarboxythiophenol | |
4-sulfanylbenzoic acid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Call a POISON CENTER or doctor/physician if
GHS Signal Word: Warning
RUO – Research Use Only