missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Mercaptobenzoic Acid 95.0+%, TCI America™

$152.25 - $152.25

Chemical Identifiers

CAS 1074-36-8
Molecular Formula C7H6O2S
Molecular Weight (g/mol) 154.18
MDL Number MFCD00016617
InChI Key LMJXSOYPAOSIPZ-UHFFFAOYSA-N
Synonym 4-mercaptobenzoic acid, 4-mercaptobenzoate, benzoic acid, 4-mercapto, 4-mercapto-benzoic acid, 4-mercapto benzoic acid, 4-thiobenzoic acid, p-mercaptobenzoic acid, benzoic acid, p-mercapto, para-mercaptobenzoate, paracarboxythiophenol
PubChem CID 95738
IUPAC Name 4-sulfanylbenzoic acid
SMILES OC(=O)C1=CC=C(S)C=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M12945G
View Documents
TCI America
M12945G
5 g
Each for $152.25
 
Description

Description

Specifications

Chemical Identifiers

1074-36-8
154.18
LMJXSOYPAOSIPZ-UHFFFAOYSA-N
95738
OC(=O)C1=CC=C(S)C=C1
C7H6O2S
MFCD00016617
4-mercaptobenzoic acid, 4-mercaptobenzoate, benzoic acid, 4-mercapto, 4-mercapto-benzoic acid, 4-mercapto benzoic acid, 4-thiobenzoic acid, p-mercaptobenzoic acid, benzoic acid, p-mercapto, para-mercaptobenzoate, paracarboxythiophenol
4-sulfanylbenzoic acid

Specifications

1074-36-8
C7H6O2S
5 g
LMJXSOYPAOSIPZ-UHFFFAOYSA-N
4-sulfanylbenzoic acid
95738
≥95.0% (GC,T)
4-Mercaptobenzoic Acid
219°C
MFCD00016617
4-mercaptobenzoic acid, 4-mercaptobenzoate, benzoic acid, 4-mercapto, 4-mercapto-benzoic acid, 4-mercapto benzoic acid, 4-thiobenzoic acid, p-mercaptobenzoic acid, benzoic acid, p-mercapto, para-mercaptobenzoate, paracarboxythiophenol
OC(=O)C1=CC=C(S)C=C1
154.18
154.18
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'