missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Methanesulfonyl)phenylboronic acid, 98+%, Thermo Scientific Chemicals

$161.46 - $161.46

Chemical Identifiers

CAS 149104-88-1
Molecular Formula C7H9BO4S
Molecular Weight (g/mol) 200.02
MDL Number MFCD01630820
InChI Key VDUKDQTYMWUSAC-UHFFFAOYSA-N
Synonym 4-methylsulfonyl phenylboronic acid, 4-methanesulfonyl phenylboronic acid, 4-methylsulfonyl phenyl boronic acid, 4-methanesulfonyl benzeneboronic acid, 4-methanesulfonylphenylboronic acid, 4-methylsulfonylphenyl boronic acid, 4-boronophenyl methyl sulphone, 4-methylsulphonyl phenylboronic acid, 4-methylsulphonyl benzeneboronic acid, 4-methanesulfonylphenyl boranediol
PubChem CID 2734364
IUPAC Name (4-methylsulfonylphenyl)boronic acid
SMILES CS(=O)(=O)C1=CC=C(C=C1)B(O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC359510010
SDS View Documents
Thermo Scientific Chemicals
359510010
1 g Glass bottle
Each for $161.46
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

149104-88-1
200.02
VDUKDQTYMWUSAC-UHFFFAOYSA-N
2734364
CS(=O)(=O)C1=CC=C(C=C1)B(O)O
C7H9BO4S
MFCD01630820
4-methylsulfonyl phenylboronic acid, 4-methanesulfonyl phenylboronic acid, 4-methylsulfonyl phenyl boronic acid, 4-methanesulfonyl benzeneboronic acid, 4-methanesulfonylphenylboronic acid, 4-methylsulfonylphenyl boronic acid, 4-boronophenyl methyl sulphone, 4-methylsulphonyl phenylboronic acid, 4-methylsulphonyl benzeneboronic acid, 4-methanesulfonylphenyl boranediol
(4-methylsulfonylphenyl)boronic acid

Specifications

149104-88-1
100.0
White
98% min. (HPLC)
C7H9BO4S
1 g
VDUKDQTYMWUSAC-UHFFFAOYSA-N
(4-methylsulfonylphenyl)boronic acid
2734364
98+%
4-(Methanesulfonyl)phenylboronic acid
98.0
275°C to 277°C
Authentic
Glass bottle
MFCD01630820
4-methylsulfonyl phenylboronic acid, 4-methanesulfonyl phenylboronic acid, 4-methylsulfonyl phenyl boronic acid, 4-methanesulfonyl benzeneboronic acid, 4-methanesulfonylphenylboronic acid, 4-methylsulfonylphenyl boronic acid, 4-boronophenyl methyl sulphone, 4-methylsulphonyl phenylboronic acid, 4-methylsulphonyl benzeneboronic acid, 4-methanesulfonylphenyl boranediol
CS(=O)(=O)C1=CC=C(C=C1)B(O)O
200.02
200.02
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only