Learn More
4-Methoxy-3-(trifluoromethyl)benzoyl chloride, 97%, Thermo Scientific Chemicals
$94.67 - $299.89
Chemical Identifiers
CAS | 98187-18-9 |
---|---|
Molecular Formula | C9H6ClF3O2 |
Molecular Weight (g/mol) | 238.59 |
MDL Number | MFCD01631576 |
InChI Key | PBKYPTQBAQQVPT-UHFFFAOYSA-N |
Synonym | 4-methoxy-3-trifluoromethyl benzoyl chloride, 3-trifluoromethyl-p-anisoyl chloride, 5-chlorocarbonyl-2-methoxybenzotrifluoride, 4methoxy-3-trifluoromethylbenzoyl chloride, 4-methoxy-5-trifluoromethylbenzoyl chloride, 3-trifluoromethyl-4-methoxy-benzoyl chloride, 4-methoxy-3-trifluoromethyl benzoylchloride, 4-methoxy-3-trifluromethyl benzoyl chloride, 4-methoxy-3-trifluoromethyl-benzoyl chloride |
PubChem CID | 2775298 |
IUPAC Name | 4-methoxy-3-(trifluoromethyl)benzoyl chloride |
SMILES | COC1=CC=C(C=C1C(F)(F)F)C(Cl)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2307203
|
Thermo Scientific Chemicals
B2307203 |
1 g |
Each for $94.67
|
|
|||||
AAB2307206
|
Thermo Scientific Chemicals
B2307206 |
5 g |
Each for $299.89
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
98187-18-9 | |
238.59 | |
PBKYPTQBAQQVPT-UHFFFAOYSA-N | |
2775298 | |
COC1=CC=C(C=C1C(F)(F)F)C(Cl)=O |
C9H6ClF3O2 | |
MFCD01631576 | |
4-methoxy-3-trifluoromethyl benzoyl chloride, 3-trifluoromethyl-p-anisoyl chloride, 5-chlorocarbonyl-2-methoxybenzotrifluoride, 4methoxy-3-trifluoromethylbenzoyl chloride, 4-methoxy-5-trifluoromethylbenzoyl chloride, 3-trifluoromethyl-4-methoxy-benzoyl chloride, 4-methoxy-3-trifluoromethyl benzoylchloride, 4-methoxy-3-trifluromethyl benzoyl chloride, 4-methoxy-3-trifluoromethyl-benzoyl chloride | |
4-methoxy-3-(trifluoromethyl)benzoyl chloride |
Specifications
98187-18-9 | |
1.515 | |
1 g | |
Moisture sensitive | |
PBKYPTQBAQQVPT-UHFFFAOYSA-N | |
4-methoxy-3-(trifluoromethyl)benzoyl chloride | |
2775298 | |
97% |
C9H6ClF3O2 | |
MFCD01631576 | |
UN3265 | |
4-methoxy-3-trifluoromethyl benzoyl chloride, 3-trifluoromethyl-p-anisoyl chloride, 5-chlorocarbonyl-2-methoxybenzotrifluoride, 4methoxy-3-trifluoromethylbenzoyl chloride, 4-methoxy-5-trifluoromethylbenzoyl chloride, 3-trifluoromethyl-4-methoxy-benzoyl chloride, 4-methoxy-3-trifluoromethyl benzoylchloride, 4-methoxy-3-trifluromethyl benzoyl chloride, 4-methoxy-3-trifluoromethyl-benzoyl chloride | |
COC1=CC=C(C=C1C(F)(F)F)C(Cl)=O | |
238.59 | |
238.59 | |
4-Methoxy-3-(trifluoromethyl)benzoyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only