Learn More
4-Methoxy-4-methyl-2-pentanone, 97%, Thermo Scientific Chemicals
$89.48 - $89.48
Chemical Identifiers
CAS | 107-70-0 |
---|---|
Molecular Formula | C7H14O2 |
Molecular Weight (g/mol) | 130.187 |
MDL Number | MFCD00043932 |
InChI Key | KOKPBCHLPVDQTK-UHFFFAOYSA-N |
Synonym | 4-methoxy-4-methyl-2-pentanone, pentoxone, pent-oxone solvent, 2-pentanone, 4-methoxy-4-methyl, 4-methyl-4-methoxy-2-pentanone, me 6 k, ccris 4840, acmc-1bwvk, dsstox_cid_5557, dsstox_rid_77828 |
PubChem CID | 7884 |
IUPAC Name | 4-methoxy-4-methylpentan-2-one |
SMILES | CC(=O)CC(C)(C)OC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2107614
|
Thermo Scientific Chemicals
B2107614 |
25 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
107-70-0 | |
130.187 | |
KOKPBCHLPVDQTK-UHFFFAOYSA-N | |
7884 | |
CC(=O)CC(C)(C)OC |
C7H14O2 | |
MFCD00043932 | |
4-methoxy-4-methyl-2-pentanone, pentoxone, pent-oxone solvent, 2-pentanone, 4-methoxy-4-methyl, 4-methyl-4-methoxy-2-pentanone, me 6 k, ccris 4840, acmc-1bwvk, dsstox_cid_5557, dsstox_rid_77828 | |
4-methoxy-4-methylpentan-2-one |
Specifications
107-70-0 | |
156°C | |
Mild | |
1.418 | |
25 g | |
4-methoxy-4-methyl-2-pentanone, pentoxone, pent-oxone solvent, 2-pentanone, 4-methoxy-4-methyl, 4-methyl-4-methoxy-2-pentanone, me 6 k, ccris 4840, acmc-1bwvk, dsstox_cid_5557, dsstox_rid_77828 | |
CC(=O)CC(C)(C)OC | |
130.187 | |
130.19 | |
4-Methoxy-4-methyl-2-pentanone |
0.91 | |
48°C (118°F) | |
C7H14O2 | |
MFCD00043932 | |
UN2293 | |
KOKPBCHLPVDQTK-UHFFFAOYSA-N | |
4-methoxy-4-methylpentan-2-one | |
7884 | |
97% |
Safety and Handling
GHS H Statement
H226-H332
Flammable liquid and vapour.
Harmful if inhaled.
P210-P233-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c
H226-H332
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: 4-METHOXY-4-METHYLPENTAN-2-ONE
EINECSNumber : 203-512-4
RTECSNumber : SA7000000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only