missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxy-4'-methyldiphenylamine 98.0+%, TCI America™
$182.68 - $549.89
Chemical Identifiers
| CAS | 39253-43-5 |
|---|---|
| Molecular Formula | C14H15NO |
| Molecular Weight (g/mol) | 213.28 |
| MDL Number | MFCD00092922 |
| InChI Key | KIDXWDVZFZMXGM-UHFFFAOYSA-N |
| Synonym | N-(4-Methoxyphenyl)-p-tolylamine |
| PubChem CID | 11435828 |
| IUPAC Name | N-(4-methoxyphenyl)-4-methylaniline |
| SMILES | COC1=CC=C(NC2=CC=C(C)C=C2)C=C1 |
Chemical Identifiers
| 39253-43-5 | |
| 213.28 | |
| KIDXWDVZFZMXGM-UHFFFAOYSA-N | |
| 11435828 | |
| COC1=CC=C(NC2=CC=C(C)C=C2)C=C1 |
| C14H15NO | |
| MFCD00092922 | |
| N-(4-Methoxyphenyl)-p-tolylamine | |
| N-(4-methoxyphenyl)-4-methylaniline |
Specifications
| 39253-43-5 | |
| C14H15NO | |
| 5 g | |
| KIDXWDVZFZMXGM-UHFFFAOYSA-N | |
| N-(4-methoxyphenyl)-4-methylaniline | |
| 11435828 | |
| ≥98.0% (GC,T) | |
| 4-Methoxy-4′-methyldiphenylamine |
| 84°C | |
| MFCD00092922 | |
| N-(4-Methoxyphenyl)-p-tolylamine | |
| COC1=CC=C(NC2=CC=C(C)C=C2)C=C1 | |
| 213.28 | |
| 213.28 | |
| Crystalline Powder |
Safety and Handling
TSCA : No