Learn More
4-Methoxy-4'-nitrobenzophenone, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2272006
Description
4-Methoxy-4'-nitrobenzophenone is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Methoxy-4′-nitrobenzophenone is used as an organic chemical synthesis intermediate.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
4-Methoxy-4'-nitrobenzophenone | |
125°C to 127°C | |
MFCD00178348 | |
4-methoxy-4'-nitrobenzophenone, 4-methoxyphenyl 4-nitrophenyl methanone, 4-methoxyphenyl-4-nitrophenyl methanone, methanone, 4-methoxyphenyl 4-nitrophenyl, 4-methoxy-phenyl-4-nitro-phenyl-methanone, acmc-1buw2, 4-methoxy-4-nitrobenzophenone, 4-methoxy-4?-nitrobenzophenone, 4-methoxy-4/'-nitrobenzophenone | |
COC1=CC=C(C=C1)C(=O)C1=CC=C(C=C1)[N+]([O-])=O | |
257.25 | |
257.25 |
1151-94-6 | |
C14H11NO4 | |
5 g | |
DXVSAELNVPXMSQ-UHFFFAOYSA-N | |
(4-methoxyphenyl)-(4-nitrophenyl)methanone | |
347772 | |
97% |
Chemical Identifiers
1151-94-6 | |
257.25 | |
DXVSAELNVPXMSQ-UHFFFAOYSA-N | |
347772 | |
COC1=CC=C(C=C1)C(=O)C1=CC=C(C=C1)[N+]([O-])=O |
C14H11NO4 | |
MFCD00178348 | |
4-methoxy-4'-nitrobenzophenone, 4-methoxyphenyl 4-nitrophenyl methanone, 4-methoxyphenyl-4-nitrophenyl methanone, methanone, 4-methoxyphenyl 4-nitrophenyl, 4-methoxy-phenyl-4-nitro-phenyl-methanone, acmc-1buw2, 4-methoxy-4-nitrobenzophenone, 4-methoxy-4?-nitrobenzophenone, 4-methoxy-4/'-nitrobenzophenone | |
(4-methoxyphenyl)-(4-nitrophenyl)methanone |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only