Learn More
4-Methoxy-o-phenylenediamine dihydrochloride, 96%, Thermo Scientific Chemicals
$181.52 - $615.36
Chemical Identifiers
CAS | 59548-39-9 |
---|---|
Molecular Formula | C7H12Cl2N2O |
Molecular Weight (g/mol) | 211.086 |
MDL Number | MFCD00052003 |
InChI Key | SXCHMHOBHJOXGC-UHFFFAOYSA-N |
Synonym | 4-methoxybenzene-1,2-diamine dihydrochloride, 4-methoxy-o-phenylenediamine dihydrochloride, 3,4-diaminoanisole dihydrochloride, 1,2-benzenediamine, 4-methoxy-, dihydrochloride, 3,4-diaminoanisole 2hcl, 4-methoxy-phenylenediamine dihydrochloride, 4-methoxy-1,2-benzenediamine dihydrochloride, 4-methoxy-benzene-1,2-diamine dihydrochloride, 4-methoxy-1,2-phenylenediamine dihydrochloride, 4-methoxy-l,2-phenylene diamine dihydrochloride |
PubChem CID | 3085161 |
IUPAC Name | 4-methoxybenzene-1,2-diamine;dihydrochloride |
SMILES | COC1=CC(=C(C=C1)N)N.Cl.Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1236306
|
Thermo Scientific Chemicals
A1236306 |
5 g |
Each for $181.52
|
|
|||||
AAA1236314
|
Thermo Scientific Chemicals
A1236314 |
25 g |
Each for $615.36
|
|
|||||
Description
4-Methoxy-o-phenylenediamine is widely used as an intermediate in organic synthesis and pharmaceuticals. It is an important precursor for the synthesis of other heterocyclic compounds. It is used in the determination of glyoxal, methylglyoxal, and diacetyl in urine using 4-methoxy-o-phenylenediamine as derivatizing reagent by High-performance liquid chromatography (HPLC).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Methoxy-o-phenylenediamine is widely used as an intermediate in organic synthesis and pharmaceuticals. It is an important precursor for the synthesis of other heterocyclic compounds. It is used in the determination of glyoxal, methylglyoxal, and diacetyl in urine using 4-methoxy-o-phenylenediamine as derivatizing reagent by High-performance liquid chromatography (HPLC).
Solubility
Soluble in water.
Notes
Store in a cool place. Incompatible with acids, acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents. Hygroscopic.
Chemical Identifiers
59548-39-9 | |
211.086 | |
SXCHMHOBHJOXGC-UHFFFAOYSA-N | |
3085161 | |
COC1=CC(=C(C=C1)N)N.Cl.Cl |
C7H12Cl2N2O | |
MFCD00052003 | |
4-methoxybenzene-1,2-diamine dihydrochloride, 4-methoxy-o-phenylenediamine dihydrochloride, 3,4-diaminoanisole dihydrochloride, 1,2-benzenediamine, 4-methoxy-, dihydrochloride, 3,4-diaminoanisole 2hcl, 4-methoxy-phenylenediamine dihydrochloride, 4-methoxy-1,2-benzenediamine dihydrochloride, 4-methoxy-benzene-1,2-diamine dihydrochloride, 4-methoxy-1,2-phenylenediamine dihydrochloride, 4-methoxy-l,2-phenylene diamine dihydrochloride | |
4-methoxybenzene-1,2-diamine;dihydrochloride |
Specifications
59548-39-9 | |
C7H12Cl2N2O | |
5 g | |
Hygroscopic | |
Soluble in water. | |
COC1=CC(=C(C=C1)N)N.Cl.Cl | |
211.086 | |
211.09 | |
4-Methoxy-o-phenylenediamine dihydrochloride |
∼219°C (decomposition) | |
MFCD00052003 | |
3699636 | |
4-methoxybenzene-1,2-diamine dihydrochloride, 4-methoxy-o-phenylenediamine dihydrochloride, 3,4-diaminoanisole dihydrochloride, 1,2-benzenediamine, 4-methoxy-, dihydrochloride, 3,4-diaminoanisole 2hcl, 4-methoxy-phenylenediamine dihydrochloride, 4-methoxy-1,2-benzenediamine dihydrochloride, 4-methoxy-benzene-1,2-diamine dihydrochloride, 4-methoxy-1,2-phenylenediamine dihydrochloride, 4-methoxy-l,2-phenylene diamine dihydrochloride | |
SXCHMHOBHJOXGC-UHFFFAOYSA-N | |
4-methoxybenzene-1,2-diamine;dihydrochloride | |
3085161 | |
≥97% |
Safety and Handling
GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 261-801-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only