Learn More
4'-Methoxyacetophenone, 99%, Thermo Scientific Chemicals
$40.93 - $226.33
Chemical Identifiers
CAS | 100-06-1 |
---|---|
Molecular Formula | C9H10O2 |
Molecular Weight (g/mol) | 150.18 |
MDL Number | MFCD00008745 |
InChI Key | NTPLXRHDUXRPNE-UHFFFAOYSA-N |
Synonym | 4'-methoxyacetophenone, 4-acetylanisole, 4-methoxyacetophenone, 1-4-methoxyphenyl ethanone, p-methoxyacetophenone, acetanisole, novatone, linarodin, vananote, ethanone, 1-4-methoxyphenyl |
PubChem CID | 7476 |
ChEBI | CHEBI:86567 |
IUPAC Name | 1-(4-methoxyphenyl)ethanone |
SMILES | COC1=CC=C(C=C1)C(C)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1116214
|
Thermo Scientific Chemicals
A1116214 |
25 g |
Each for $40.93
|
|
|||||
AAA1116230
|
Thermo Scientific Chemicals
A1116230 |
250 g |
Each for $77.40
|
|
|||||
AAA111620B
|
Thermo Scientific Chemicals
A111620B |
1000 g |
Each for $226.33
|
|
|||||
Description
4'-Methoxyacetophenone is used as a component of perfumes and as chemical intermediate in the manufacture of pharmaceuticals, resins, flavouring agents. And also used to study ruthenium catalyzed step growth copolymerization of 4?-methoxyacetophenone with α,ω-dienes to yield copolymers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4′-Methoxyacetophenone is used as a component of perfumes and as chemical intermediate in the manufacture of pharmaceuticals, resins, flavouring agents. And also used to study ruthenium catalyzed step growth copolymerization of 4′-methoxyacetophenone with α,ω-dienes to yield copolymers.
Solubility
Insoluble in water.
Notes
Light Sensitive, Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
100-06-1 | |
150.18 | |
NTPLXRHDUXRPNE-UHFFFAOYSA-N | |
7476 | |
1-(4-methoxyphenyl)ethanone |
C9H10O2 | |
MFCD00008745 | |
4'-methoxyacetophenone, 4-acetylanisole, 4-methoxyacetophenone, 1-4-methoxyphenyl ethanone, p-methoxyacetophenone, acetanisole, novatone, linarodin, vananote, ethanone, 1-4-methoxyphenyl | |
CHEBI:86567 | |
COC1=CC=C(C=C1)C(C)=O |
Specifications
100-06-1 | |
1.08 | |
138°C (280°F) | |
C9H10O2 | |
25 g | |
Light sensitive | |
Insoluble in water. | |
COC1=CC=C(C=C1)C(C)=O | |
150.18 | |
CHEBI:86567 | |
99% |
36°C to 40°C | |
258°C to 260°C | |
Characteristic | |
MFCD00008745 | |
742313 | |
4'-methoxyacetophenone, 4-acetylanisole, 4-methoxyacetophenone, 1-4-methoxyphenyl ethanone, p-methoxyacetophenone, acetanisole, novatone, linarodin, vananote, ethanone, 1-4-methoxyphenyl | |
NTPLXRHDUXRPNE-UHFFFAOYSA-N | |
1-(4-methoxyphenyl)ethanone | |
7476 | |
150.18 | |
4'-Methoxyacetophenone |
Safety and Handling
GHS H Statement
H301-H315
Toxic if swallowed.
Causes skin irritation.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 202-815-9
RTECSNumber : AM9240000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only