Learn More
4-Methoxybenzenesulfonamide, 98%, Thermo Scientific Chemicals
$127.56 - $387.34
Chemical Identifiers
CAS | 1129-26-6 |
---|---|
Molecular Formula | C7H9NO3S |
Molecular Weight (g/mol) | 187.213 |
MDL Number | MFCD00025392 |
InChI Key | MSFQEZBRFPAFEX-UHFFFAOYSA-N |
Synonym | 4-methoxybenzenesulphonamide, p-methoxybenzenesulfonamide, 4-methoxybenzene-1-sulfonamide, benzenesulfonamide, 4-methoxy, 4-methoxy-benzenesulfonamide, benzenesulfonamide, p-methoxy, benzenesulfonamide, p-methoxy-6ci,7ci,8ci, 3lka, p-anisolesulfonamide, acmc-1bvgf |
PubChem CID | 70789 |
IUPAC Name | 4-methoxybenzenesulfonamide |
SMILES | COC1=CC=C(C=C1)S(=O)(=O)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0896106
|
Thermo Scientific Chemicals
L0896106 |
5 g |
Each for $127.56
|
|
|||||
AAL0896114
|
Thermo Scientific Chemicals
L0896114 |
25 g |
Each for $387.34
|
|
|||||
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Slightly soluble in water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
1129-26-6 | |
187.213 | |
MSFQEZBRFPAFEX-UHFFFAOYSA-N | |
70789 | |
COC1=CC=C(C=C1)S(=O)(=O)N |
C7H9NO3S | |
MFCD00025392 | |
4-methoxybenzenesulphonamide, p-methoxybenzenesulfonamide, 4-methoxybenzene-1-sulfonamide, benzenesulfonamide, 4-methoxy, 4-methoxy-benzenesulfonamide, benzenesulfonamide, p-methoxy, benzenesulfonamide, p-methoxy-6ci,7ci,8ci, 3lka, p-anisolesulfonamide, acmc-1bvgf | |
4-methoxybenzenesulfonamide |
Specifications
1129-26-6 | |
C7H9NO3S | |
5 g | |
4-methoxybenzenesulphonamide, p-methoxybenzenesulfonamide, 4-methoxybenzene-1-sulfonamide, benzenesulfonamide, 4-methoxy, 4-methoxy-benzenesulfonamide, benzenesulfonamide, p-methoxy, benzenesulfonamide, p-methoxy-6ci,7ci,8ci, 3lka, p-anisolesulfonamide, acmc-1bvgf | |
MSFQEZBRFPAFEX-UHFFFAOYSA-N | |
4-methoxybenzenesulfonamide | |
70789 | |
98% |
110°C to 114°C | |
MFCD00025392 | |
2047881 | |
Slightly soluble in water. | |
COC1=CC=C(C=C1)S(=O)(=O)N | |
187.213 | |
187.22 | |
4-Methoxybenzenesulfonamide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
RTECSNumber : DB2900000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only