Learn More
4-Methoxybenzophenone, 98+%, Thermo Scientific Chemicals
$114.14 - $288.53
Chemical Identifiers
CAS | 611-94-9 |
---|---|
Molecular Formula | C14H12O2 |
Molecular Weight (g/mol) | 212.248 |
MDL Number | MFCD00008403 |
InChI Key | SWFHGTMLYIBPPA-UHFFFAOYSA-N |
Synonym | 4-methoxybenzophenone, 4-methoxyphenyl phenyl methanone, p-methoxybenzophenone, benzophenone, 4-methoxy, methanone, 4-methoxyphenyl phenyl, phenyl p-anisyl ketone, 4-benzoylanisole, 4-methoxyphenyl phenylmethanone, unii-i4xj07373m, 4-methoxyphenyl-phenylmethanone |
PubChem CID | 69146 |
IUPAC Name | (4-methoxyphenyl)-phenylmethanone |
SMILES | COC1=CC=C(C=C1)C(=O)C2=CC=CC=C2 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2312314
|
Thermo Scientific Chemicals
B2312314 |
25 g |
Each for $114.14
|
|
|||||
AAB2312322
|
Thermo Scientific Chemicals
B2312322 |
100 g |
Each for $288.53
|
|
|||||
Description
4-Methoxybenzophenone and its analogues showed diverse plant growth-regulating actions such as inhibition of shoot and root growth, induction of chlorosis, and a disturbance in phototropism or geotropism. It is also used as a photo polymerization catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Methoxybenzophenone and its analogues showed diverse plant growth-regulating actions such as inhibition of shoot and root growth, induction of chlorosis, and a disturbance in phototropism or geotropism. It is also used as a photo polymerization catalyst.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agent.
Chemical Identifiers
611-94-9 | |
212.248 | |
SWFHGTMLYIBPPA-UHFFFAOYSA-N | |
69146 | |
COC1=CC=C(C=C1)C(=O)C2=CC=CC=C2 |
C14H12O2 | |
MFCD00008403 | |
4-methoxybenzophenone, 4-methoxyphenyl phenyl methanone, p-methoxybenzophenone, benzophenone, 4-methoxy, methanone, 4-methoxyphenyl phenyl, phenyl p-anisyl ketone, 4-benzoylanisole, 4-methoxyphenyl phenylmethanone, unii-i4xj07373m, 4-methoxyphenyl-phenylmethanone | |
(4-methoxyphenyl)-phenylmethanone |
Specifications
611-94-9 | |
354°C to 356°C | |
MFCD00008403 | |
1104713 | |
Insoluble in water. | |
COC1=CC=C(C=C1)C(=O)C2=CC=CC=C2 | |
212.248 | |
212.25 | |
4-Methoxybenzophenone |
60°C to 63°C | |
C14H12O2 | |
25 g | |
4-methoxybenzophenone, 4-methoxyphenyl phenyl methanone, p-methoxybenzophenone, benzophenone, 4-methoxy, methanone, 4-methoxyphenyl phenyl, phenyl p-anisyl ketone, 4-benzoylanisole, 4-methoxyphenyl phenylmethanone, unii-i4xj07373m, 4-methoxyphenyl-phenylmethanone | |
SWFHGTMLYIBPPA-UHFFFAOYSA-N | |
(4-methoxyphenyl)-phenylmethanone | |
69146 | |
≥98% |
Safety and Handling
EINECSNumber : 210-285-5
RTECSNumber : PC4962500
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only