missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methoxybenzyl isocyanate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 319400010

 View more versions of this product

Catalog No. AC319400010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Methoxybenzyl isocyanate
98.5
90.0°C (0.6 mmHg)
Glass bottle
1.5305 to 1.5325
1 g
QRBHVARIMDDOOV-UHFFFAOYSA-N
1-(isocyanatomethyl)-4-methoxybenzene
187910
99%
56651-60-6
100.0
99%
C9H9NO2
MFCD00673064
4-methoxybenzyl isocyanate, 1-isocyanatomethyl-4-methoxybenzene, p-methoxybenzyl isocyanate, 4-methoxybenzylisocyanate, isocyanic acid, p-methoxybenzyl, benzene, 1-isocyanatomethyl-4-methoxy, 4-methoxyphenyl methanisocyanate, acmc-20amf5, p-methoxybenzylisocyanate, p-metoxybenzyl-isocyanate
COC1=CC=C(CN=C=O)C=C1
163.18
163.18

Chemical Identifiers

56651-60-6
163.18
QRBHVARIMDDOOV-UHFFFAOYSA-N
187910
COC1=CC=C(CN=C=O)C=C1
C9H9NO2
MFCD00673064
4-methoxybenzyl isocyanate, 1-isocyanatomethyl-4-methoxybenzene, p-methoxybenzyl isocyanate, 4-methoxybenzylisocyanate, isocyanic acid, p-methoxybenzyl, benzene, 1-isocyanatomethyl-4-methoxy, 4-methoxyphenyl methanisocyanate, acmc-20amf5, p-methoxybenzylisocyanate, p-metoxybenzyl-isocyanate
1-(isocyanatomethyl)-4-methoxybenzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only