Learn More
4-Methoxybenzyl mercaptan, 98%, Thermo Scientific Chemicals
$105.84 - $321.47
Chemical Identifiers
CAS | 6258-60-2 |
---|---|
Molecular Formula | C8H10OS |
Molecular Weight (g/mol) | 154.23 |
MDL Number | MFCD00004871 |
InChI Key | PTDVPWWJRCOIIO-UHFFFAOYSA-N |
Synonym | 4-methoxybenzyl mercaptan, 4-methoxy-alpha-toluenethiol, 4-methoxybenzylmercaptan, 4-methoxyphenyl methanethiol, 4-methoxy benzylmercaptan, 4-methoxybenzenemethanethiol, benzenemethanethiol, 4-methoxy, p-methoxybenzyl mercaptan, p-methoxybenzylthiol, p-methoxybenzylmercaptan |
PubChem CID | 80407 |
IUPAC Name | (4-methoxyphenyl)methanethiol |
SMILES | COC1=CC=C(CS)C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2254206
|
Thermo Scientific Chemicals
B2254206 |
5 g |
Each for $105.84
|
|
|||||
AAB2254214
|
Thermo Scientific Chemicals
B2254214 |
25 g |
Each for $321.47
|
|
|||||
Description
4-Methoxybenzyl mercaptan is used to synthesize many ω-mercapto amino acids with side-chain lengths ranging from 3-5 methylene units. It is used as an internal standard in the Quantitative determination of wine polyfunctional mercaptans at the level of nanogram per liter.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Methoxybenzyl mercaptan is used to synthesize many ω-mercapto amino acids with side-chain lengths ranging from 3-5 methylene units. It is used as an internal standard in the Quantitative determination of wine polyfunctional mercaptans at the level of nanogram per liter.
Solubility
Not miscible or difficult to mix in water.
Notes
Air sensitive. Store under inert gas. Incompatible with oxidizing agents, acids, bases, air.
Chemical Identifiers
6258-60-2 | |
154.23 | |
PTDVPWWJRCOIIO-UHFFFAOYSA-N | |
80407 | |
COC1=CC=C(CS)C=C1 |
C8H10OS | |
MFCD00004871 | |
4-methoxybenzyl mercaptan, 4-methoxy-alpha-toluenethiol, 4-methoxybenzylmercaptan, 4-methoxyphenyl methanethiol, 4-methoxy benzylmercaptan, 4-methoxybenzenemethanethiol, benzenemethanethiol, 4-methoxy, p-methoxybenzyl mercaptan, p-methoxybenzylthiol, p-methoxybenzylmercaptan | |
(4-methoxyphenyl)methanethiol |
Specifications
6258-60-2 | |
90°C to 95°C (0.5 mmHg) | |
Unpleasant | |
1.5755 | |
5 g | |
Air Sensitive | |
Not miscible or difficult to mix in water. | |
COC1=CC=C(CS)C=C1 | |
154.23 | |
154.23 | |
4-Methoxybenzyl mercaptan |
1.112 | |
>110°C (230°F) | |
C8H10OS | |
MFCD00004871 | |
471686 | |
4-methoxybenzyl mercaptan, 4-methoxy-alpha-toluenethiol, 4-methoxybenzylmercaptan, 4-methoxyphenyl methanethiol, 4-methoxy benzylmercaptan, 4-methoxybenzenemethanethiol, benzenemethanethiol, 4-methoxy, p-methoxybenzyl mercaptan, p-methoxybenzylthiol, p-methoxybenzylmercaptan | |
PTDVPWWJRCOIIO-UHFFFAOYSA-N | |
(4-methoxyphenyl)methanethiol | |
80407 | |
98% |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
H317-H334
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 228-393-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only