missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Methoxycarbonylamino)benzeneboronic acid, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H53027MD

 View more versions of this product

Catalog No. AAH53027MD


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-(Methoxycarbonylamino)benzeneboronic acid
234°C to 236°C
MFCD03788425
4-methoxycarbonylamino phenylboronic acid, 4-methoxycarbonyl amino phenyl boronic acid, 4-methoxycarbonylamino benzeneboronic acid, 4-methoxycarbonyl amino phenylboronic acid, 4-methoxycarbonylamino phenyl boronic acid, acmc-209q0y, 4-methoxycarbonylamino-phenyl boronic acid, 4-methoxycarbonylamino benzene boronic acid, 4-methoxycarbonyl amino phenyl boronicacid
B(C1=CC=C(C=C1)NC(=O)OC)(O)O
194.981
194.98
850567-96-3
C8H10BNO4
250 mg
ROAKVYQVTMOEGR-UHFFFAOYSA-N
[4-(methoxycarbonylamino)phenyl]boronic acid
2773494
97%

Chemical Identifiers

850567-96-3
194.981
ROAKVYQVTMOEGR-UHFFFAOYSA-N
2773494
B(C1=CC=C(C=C1)NC(=O)OC)(O)O
C8H10BNO4
MFCD03788425
4-methoxycarbonylamino phenylboronic acid, 4-methoxycarbonyl amino phenyl boronic acid, 4-methoxycarbonylamino benzeneboronic acid, 4-methoxycarbonyl amino phenylboronic acid, 4-methoxycarbonylamino phenyl boronic acid, acmc-209q0y, 4-methoxycarbonylamino-phenyl boronic acid, 4-methoxycarbonylamino benzene boronic acid, 4-methoxycarbonyl amino phenyl boronicacid
[4-(methoxycarbonylamino)phenyl]boronic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only