missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Methoxyphenylacetic Anhydride 98.0+%, TCI America™
Supplier: TCI America M096825G
Specifications
4-Methoxyphenylacetic Anhydride | |
76°C | |
C18H18O5 | |
25 g | |
COC1=CC=C(C=C1)CC(=O)OC(=O)CC2=CC=C(C=C2)OC | |
314.34 | |
314.34 | |
Crystalline Powder |
3951-10-8 | |
White-Yellow | |
MFCD00059089 | |
KYCAEIXHUXBNTQ-UHFFFAOYSA-N | |
[2-(4-methoxyphenyl)acetyl] 2-(4-methoxyphenyl)acetate | |
14086930 | |
≥98.0% (T) |
Chemical Identifiers
3951-10-8 | |
314.34 | |
KYCAEIXHUXBNTQ-UHFFFAOYSA-N | |
[2-(4-methoxyphenyl)acetyl] 2-(4-methoxyphenyl)acetate |
C18H18O5 | |
MFCD00059089 | |
14086930 | |
COC1=CC=C(C=C1)CC(=O)OC(=O)CC2=CC=C(C=C2)OC |
Safety and Handling
TSCA : No