Learn More
4-Methoxyphenylacetylene, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H2628306
Description
4-Ethynylanisole was used in the synthesis of photo luminescent 1,2-dihydrophosphinines via a [4 + 2] cycloaddition. Along with an arylboronic acid and sodium azide in a copper-catalyzed, three-component synthesis of trisubstituted 1,2,4-triazoles. In a study of a gold (III)-catalyzed hydroamination of alkynes leading to N-vinylindoles3.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Ethynylanisole was used in the synthesis of photo luminescent 1,2-dihydrophosphinines via a [4 + 2] cycloaddition. Along with an arylboronic acid and sodium azide in a copper-catalyzed, three-component synthesis of trisubstituted 1,2,4-triazoles. In a study of a gold (III)-catalyzed hydroamination of alkynes leading to N-vinylindoles3.
Solubility
Insoluble in water. Soluble in chloroform, acetone, dichloromethane, and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
4-Methoxyphenylacetylene | |
27°C to 29°C | |
87°C to 91°C (11 mmHg) | |
C9H8O | |
MFCD00168815 | |
4-ethynylanisole, 4-methoxyphenylacetylene, 4'-methoxyphenyl acetylene, 1-ethynyl-4-methoxy-benzene, 1-eth-1-ynyl-4-methoxybenzene, benzene, 1-ethynyl-4-methoxy, 4-methoxyphenyl acetylene, 4-ethynyl-1-methoxybenzene, 4'-methoxyphenylacetylene, p-ethynylanisole | |
KBIAVTUACPKPFJ-UHFFFAOYSA-N | |
1-ethynyl-4-methoxybenzene | |
251020 | |
99% |
768-60-5 | |
1.019 | |
82°C (180°F) | |
1.563 | |
5 g | |
Insoluble in water. Soluble in chloroform,acetone,dichloromethane,and methanol. | |
COC1=CC=C(C=C1)C#C | |
132.162 | |
132.16 |
Chemical Identifiers
768-60-5 | |
132.162 | |
KBIAVTUACPKPFJ-UHFFFAOYSA-N | |
251020 | |
COC1=CC=C(C=C1)C#C |
C9H8O | |
MFCD00168815 | |
4-ethynylanisole, 4-methoxyphenylacetylene, 4'-methoxyphenyl acetylene, 1-ethynyl-4-methoxy-benzene, 1-eth-1-ynyl-4-methoxybenzene, benzene, 1-ethynyl-4-methoxy, 4-methoxyphenyl acetylene, 4-ethynyl-1-methoxybenzene, 4'-methoxyphenylacetylene, p-ethynylanisole | |
1-ethynyl-4-methoxybenzene |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335-H500
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only