missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methoxyphenylboronic acid, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 309480050

 View more versions of this product

Catalog No. AC309480050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Methoxyphenylboronic acid
96.0
208.0°C to 210.0°C
Authentic
Glass bottle
CH3OC6H4B(OH)2
5 g
VOAAEKKFGLPLLU-UHFFFAOYSA-N
(4-methoxyphenyl)boronic acid
201262
97%
5720-07-0
100.0
White to Beige
97%
C7H9BO3
MFCD00039139
4-methoxyphenyl boronic acid, 4-methoxybenzeneboronic acid, p-anisylboronic acid, p-methoxyphenylboronic acid, p-methoxybenzeneboronic acid, 4-methoxyphenyl boranediol, benzeneboronic acid, p-methoxy, 4-boronoanisole, 4-methoxyphenylboronicacid
COC1=CC=C(C=C1)B(O)O
151.96
151.96
Crystalline Powder

Chemical Identifiers

5720-07-0
151.96
VOAAEKKFGLPLLU-UHFFFAOYSA-N
201262
COC1=CC=C(C=C1)B(O)O
C7H9BO3
MFCD00039139
4-methoxyphenyl boronic acid, 4-methoxybenzeneboronic acid, p-anisylboronic acid, p-methoxyphenylboronic acid, p-methoxybenzeneboronic acid, 4-methoxyphenyl boranediol, benzeneboronic acid, p-methoxy, 4-boronoanisole, 4-methoxyphenylboronicacid
(4-methoxyphenyl)boronic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only