missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methyl-1,2,3-thiadiazole-5-carboxylic acid, 97%, Thermo Scientific™

$18.23 - $18.23

Chemical Identifiers

CAS 18212-21-0
Molecular Formula C4H3N2O2S
Molecular Weight (g/mol) 143.14
MDL Number MFCD00173830
InChI Key NHHQOYLPBUYHQU-UHFFFAOYSA-M
Synonym 4-methyl-1,2,3-thiadiazole-5-carboxylic acid, 4-methyl-1,2,3 thiadiazole-5-carboxylic acid, 1,2,3-thiadiazole-5-carboxylic acid, 4-methyl, pubchem10092, acmc-209xfo, 4-methyl-1,2,3-thiadiazole-5-carboxylicacid, tos-bb-0050, 4-methyl-1,2,3 thiadiazol-5-carboxylic acid, 4-methyl-5-hydroxycarbonyl-1,2,3-thiadiazole, 4methyl-1,2,3 thiadiazole-5-carboxylic acid
PubChem CID 1381992
IUPAC Name 4-methylthiadiazole-5-carboxylic acid
SMILES CC1=C(SN=N1)C([O-])=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC02301DA
SDS View Documents
Thermo Scientific Chemicals
CC02301DA
1 g
Each for $18.23
 
Description

Description

Specifications

Chemical Identifiers

18212-21-0
143.14
NHHQOYLPBUYHQU-UHFFFAOYSA-M
1381992
CC1=C(SN=N1)C([O-])=O
C4H3N2O2S
MFCD00173830
4-methyl-1,2,3-thiadiazole-5-carboxylic acid, 4-methyl-1,2,3 thiadiazole-5-carboxylic acid, 1,2,3-thiadiazole-5-carboxylic acid, 4-methyl, pubchem10092, acmc-209xfo, 4-methyl-1,2,3-thiadiazole-5-carboxylicacid, tos-bb-0050, 4-methyl-1,2,3 thiadiazol-5-carboxylic acid, 4-methyl-5-hydroxycarbonyl-1,2,3-thiadiazole, 4methyl-1,2,3 thiadiazole-5-carboxylic acid
4-methylthiadiazole-5-carboxylic acid

Specifications

18212-21-0
100.0
Amber glass bottle
MFCD00173830
4-methyl-1,2,3-thiadiazole-5-carboxylic acid, 4-methyl-1,2,3 thiadiazole-5-carboxylic acid, 1,2,3-thiadiazole-5-carboxylic acid, 4-methyl, pubchem10092, acmc-209xfo, 4-methyl-1,2,3-thiadiazole-5-carboxylicacid, tos-bb-0050, 4-methyl-1,2,3 thiadiazol-5-carboxylic acid, 4-methyl-5-hydroxycarbonyl-1,2,3-thiadiazole, 4methyl-1,2,3 thiadiazole-5-carboxylic acid
CC1=C(SN=N1)C([O-])=O
143.14
144.15
4-Methyl-1, 2, 3-thiadiazole-5-carboxylic acid
96.5
97%
C4H3N2O2S
1 g
NHHQOYLPBUYHQU-UHFFFAOYSA-M
4-methylthiadiazole-5-carboxylic acid
1381992
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'