missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methyl-1-pentanol 99.0+%, TCI America™

Supplier:  TCI America M077525ML

 View more versions of this product

Catalog No. M077525ML


Explore available promotions

Description

Description

Specifications

Specifications

4-Methyl-1-pentanol
Colorless
C6H14O
25 mL
4-methyl-1-pentanol, isohexanol, isohexyl alcohol, 4-methylpentanol, iso-hexanol, 2-methyl-5-pentanol, 1-pentanol, 4-methyl, pentanol, 4-methyl, 4-methyl-pentan-1-ol, unii-x796xfp7d4
CC(C)CCCO
102.177
CHEBI:63910
≥99.0% (GC)
626-89-1
152°C
MFCD00002962
1987
PCWGTDULNUVNBN-UHFFFAOYSA-N
4-methylpentan-1-ol
12296
102.18
Liquid

Chemical Identifiers

626-89-1
102.177
PCWGTDULNUVNBN-UHFFFAOYSA-N
12296
4-methylpentan-1-ol
C6H14O
MFCD00002962
4-methyl-1-pentanol, isohexanol, isohexyl alcohol, 4-methylpentanol, iso-hexanol, 2-methyl-5-pentanol, 1-pentanol, 4-methyl, pentanol, 4-methyl, 4-methyl-pentan-1-ol, unii-x796xfp7d4
CHEBI:63910
CC(C)CCCO
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0217

RTECSNumber : NR3020000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions