missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methyl-2-nitrophenol, 97%, Thermo Scientific Chemicals

$96.41 - $1208.93

Chemical Identifiers

CAS 119-33-5
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD00007120
InChI Key SYDNSSSQVSOXTN-UHFFFAOYSA-N
Synonym 2-nitro-p-cresol, phenol, 4-methyl-2-nitro, o-nitro-p-cresol, 2-nitro-4-methylphenol, 4-hydroxy-3-nitrotoluene, p-cresol, 2-nitro, 2-nitro-4-cresol, unii-p92kpk2nl3, 4-methyl-2-nitro-phenol, p92kpk2nl3
PubChem CID 8391
IUPAC Name 4-methyl-2-nitrophenol
SMILES CC1=CC(=C(C=C1)O)[N+](=O)[O-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1200914
View Documents
Thermo Scientific Chemicals
A1200914
25 g
Each for $96.41
 
AAA1200922
View Documents
Thermo Scientific Chemicals
A1200922
100 g
Each for $306.79
 
AAA1200936
View Documents
Thermo Scientific Chemicals
A1200936
500 g
Each for $1,208.93
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

119-33-5
153.137
SYDNSSSQVSOXTN-UHFFFAOYSA-N
8391
CC1=CC(=C(C=C1)O)[N+](=O)[O-]
C7H7NO3
MFCD00007120
2-nitro-p-cresol, phenol, 4-methyl-2-nitro, o-nitro-p-cresol, 2-nitro-4-methylphenol, 4-hydroxy-3-nitrotoluene, p-cresol, 2-nitro, 2-nitro-4-cresol, unii-p92kpk2nl3, 4-methyl-2-nitro-phenol, p92kpk2nl3
4-methyl-2-nitrophenol

Specifications

119-33-5
1.24
108°C (226°F)
C7H7NO3
25 g
2-nitro-p-cresol, phenol, 4-methyl-2-nitro, o-nitro-p-cresol, 2-nitro-4-methylphenol, 4-hydroxy-3-nitrotoluene, p-cresol, 2-nitro, 2-nitro-4-cresol, unii-p92kpk2nl3, 4-methyl-2-nitro-phenol, p92kpk2nl3
CC1=CC(=C(C=C1)O)[N+](=O)[O-]
153.137
153.14
4-Methyl-2-nitrophenol
32°C to 35°C
125°C (22 mmHg)
Phenol-like
MFCD00007120
1868022
SYDNSSSQVSOXTN-UHFFFAOYSA-N
4-methyl-2-nitrophenol
8391
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H312-H315-H319-H335
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 204-315-6

RTECSNumber : GP2800000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only