Learn More
4-Methyl-2-pentanol, 99+%
CAS: 108-11-2 | C6H14O | 102.18 g/mol
$57.55 - $168.31
Chemical Identifiers
| CAS | 108-11-2 |
|---|---|
| Molecular Formula | C6H14O |
| Molecular Weight (g/mol) | 102.18 |
| MDL Number | MFCD00004550 |
| InChI Key | WVYWICLMDOOCFB-UHFFFAOYSA-N |
| Synonym | 4-methyl-2-pentanol, 2-pentanol, 4-methyl, isobutylmethylcarbinol, 2-methyl-4-pentanol, methyl amyl alcohol, methyl isobutyl carbinol, mibc, 1,3-dimethylbutanol, 4-methylpentanol-2, isobutylmethylmethanol |
| PubChem CID | 7910 |
| IUPAC Name | 4-methylpentan-2-ol |
| SMILES | CC(C)CC(C)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC149380050
|
Thermo Scientific Chemicals
149380050 |
5 mL | Glass bottle |
Each for $57.55
|
|
||||
|
AC149385000
|
Thermo Scientific Chemicals
149385000 |
500 mL | Glass bottle |
Each for $58.34
|
|
||||
|
AC149380010
|
Thermo Scientific Chemicals
149380010 |
1 L | Glass bottle |
Each for $84.75
Case of 4 Each for $0.00
|
|
||||
|
AC149380025
|
Thermo Scientific Chemicals
149380025 |
2.5 L | Glass bottle |
Each for $168.31
Case of 4 Each for $0.00
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 108-11-2 | |
| 102.18 | |
| WVYWICLMDOOCFB-UHFFFAOYSA-N | |
| 7910 | |
| CC(C)CC(C)O |
| C6H14O | |
| MFCD00004550 | |
| 4-methyl-2-pentanol, 2-pentanol, 4-methyl, isobutylmethylcarbinol, 2-methyl-4-pentanol, methyl amyl alcohol, methyl isobutyl carbinol, mibc, 1,3-dimethylbutanol, 4-methylpentanol-2, isobutylmethylmethanol | |
| 4-methylpentan-2-ol |
Specifications
| 108-11-2 | |
| 100.0 | |
| Colorless | |
| 132.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4100 to 1.4120 | |
| MFCD00004550 | |
| 01, 410 | |
| 4-methyl-2-pentanol, 2-pentanol, 4-methyl, isobutylmethylcarbinol, 2-methyl-4-pentanol, methyl amyl alcohol, methyl isobutyl carbinol, mibc, 1,3-dimethylbutanol, 4-methylpentanol-2, isobutylmethylmethanol | |
| WVYWICLMDOOCFB-UHFFFAOYSA-N | |
| 4-methylpentan-2-ol | |
| 7910 | |
| 99+% | |
| 4-Methyl-2-pentanol |
| 99.0 | |
| -90.0°C | |
| 0.8000g/mL | |
| 41°C | |
| 99+% | |
| C6H14O | |
| (CH3)2CHCH2CH(OH)CH3 | |
| 5 mL | |
| 0.8 | |
| Solubility in water: 20g/L. Other solubilities: misscible with most common organic solvents | |
| CC(C)CC(C)O | |
| 102.18 | |
| 102.18 | |
| Liquid |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Flammable liquid and vapor.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 203-551-7
RUO – Research Use Only