missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methyl-2-pyridyl Trifluoromethanesulfonate 97.0+%, TCI America™

$86.90 - $428.57

Chemical Identifiers

CAS 179260-78-7
Molecular Formula C7H6F3NO3S
Molecular Weight (g/mol) 241.184
InChI Key AZUJRVRRUXHHDV-UHFFFAOYSA-N
Synonym Trifluoromethanesulfonic Acid 4-Methyl-2-pyridyl Ester
PubChem CID 10857619
IUPAC Name (4-methylpyridin-2-yl) trifluoromethanesulfonate
SMILES CC1=CC(=NC=C1)OS(=O)(=O)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M19741G
View Documents
TCI America
M19741G
1 g
Each for $86.90
 
M19745G
View Documents
TCI America
M19745G
5 g
Each for $428.57
 
Description

Description

Specifications

Chemical Identifiers

179260-78-7
241.184
Trifluoromethanesulfonic Acid 4-Methyl-2-pyridyl Ester
(4-methylpyridin-2-yl) trifluoromethanesulfonate
C7H6F3NO3S
AZUJRVRRUXHHDV-UHFFFAOYSA-N
10857619
CC1=CC(=NC=C1)OS(=O)(=O)C(F)(F)F

Specifications

179260-78-7
C7H6F3NO3S
Trifluoromethanesulfonic Acid 4-Methyl-2-pyridyl Ester
CC1=CC(=NC=C1)OS(=O)(=O)C(F)(F)F
241.184
241.18
Liquid
Yellow
1 g
AZUJRVRRUXHHDV-UHFFFAOYSA-N
(4-methylpyridin-2-yl) trifluoromethanesulfonate
10857619
≥97.0% (GC)
4-Methyl-2-pyridyl Trifluoromethanesulfonate
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
missing translation for 'documents'

missing translation for 'documents'