missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methyl-2-thiazolecarboxaldehyde, 97%, Thermo Scientific™

Chemical Identifiers

CAS 13750-68-0
Molecular Formula C5H5NOS
Molecular Weight (g/mol) 127.17
MDL Number MFCD00022450
InChI Key NYMCQLLAIMUVSY-UHFFFAOYSA-N
Synonym 4-methylthiazole-2-carbaldehyde, 4-methyl-2-thiazolecarboxaldehyde, 4-methyl-thiazole-2-carbaldehyde, 4-methyl-1,3-thiazole-2-carboxaldehyde, 2-thiazolecarboxaldehyde, 4-methyl, 4-methylthiazole-2-carboxaldehyde, 4-methyl-2-formylthiazole, 2-formyl-4-methyl-1,3-thiazole, 4-methyl-2-thiazole carboxaldehyde, pubchem15253
PubChem CID 4263432
IUPAC Name 4-methyl-1,3-thiazole-2-carbaldehyde
SMILES CC1=CSC(=N1)C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC429350010
SDS View Documents
Thermo Scientific Chemicals
429350010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC429350050
SDS View Documents
Thermo Scientific Chemicals
429350050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

13750-68-0
127.17
NYMCQLLAIMUVSY-UHFFFAOYSA-N
4263432
CC1=CSC(=N1)C=O
C5H5NOS
MFCD00022450
4-methylthiazole-2-carbaldehyde, 4-methyl-2-thiazolecarboxaldehyde, 4-methyl-thiazole-2-carbaldehyde, 4-methyl-1,3-thiazole-2-carboxaldehyde, 2-thiazolecarboxaldehyde, 4-methyl, 4-methylthiazole-2-carboxaldehyde, 4-methyl-2-formylthiazole, 2-formyl-4-methyl-1,3-thiazole, 4-methyl-2-thiazole carboxaldehyde, pubchem15253
4-methyl-1,3-thiazole-2-carbaldehyde

Specifications

13750-68-0
100.0
67°C
Glass bottle
MFCD00022450
4-methylthiazole-2-carbaldehyde, 4-methyl-2-thiazolecarboxaldehyde, 4-methyl-thiazole-2-carbaldehyde, 4-methyl-1,3-thiazole-2-carboxaldehyde, 2-thiazolecarboxaldehyde, 4-methyl, 4-methylthiazole-2-carboxaldehyde, 4-methyl-2-formylthiazole, 2-formyl-4-methyl-1,3-thiazole, 4-methyl-2-thiazole carboxaldehyde, pubchem15253
CC1=CSC(=N1)C=O
127.17
127.17
4-Methyl-2-thiazolecarboxaldehyde
96.0
90.0°C to 92.0°C (12.0 mbar)
96% min. (GC)
C5H5NOS
1g
NYMCQLLAIMUVSY-UHFFFAOYSA-N
4-methyl-1,3-thiazole-2-carbaldehyde
4263432
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only