Learn More
4-Methyl-2-(trifluoromethyl)benzoic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H3253403
Description
4-Methyl-2-(trifluoromethyl)benzoic acid is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Methyl-2-(trifluoromethyl)benzoic acid is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed co
Specifications
4-Methyl-2-(trifluoromethyl)benzoic acid | |
C9H7F3O2 | |
1 g | |
Insoluble in water. | |
CC1=CC(=C(C=C1)C(O)=O)C(F)(F)F | |
204.15 | |
204.15 |
120985-64-0 | |
MFCD01631591 | |
4-methyl-2-trifluoromethyl benzoic acid, 2-trifluoromethy-4-methybenzoic acid, 2-trifluoromethyl-4-methyl-benzoic acid, 4-methyl-2-trifluoromethyl-benzoic acid, 2-carboxy-5-methylbenzotrifluoride, 2-trifluoromethyl-p-toluic acid, 4-carboxy-3-trifluoromethyl toluene, 2-trifluoromethyl-4-methyl benzoic acid, 4-methyl-2-trifluoromethyl benzoicacid | |
PHJOGJTZXVBKJO-UHFFFAOYSA-N | |
4-methyl-2-(trifluoromethyl)benzoic acid | |
2775591 | |
98% |
Chemical Identifiers
120985-64-0 | |
204.15 | |
PHJOGJTZXVBKJO-UHFFFAOYSA-N | |
2775591 | |
CC1=CC(=C(C=C1)C(O)=O)C(F)(F)F |
C9H7F3O2 | |
MFCD01631591 | |
4-methyl-2-trifluoromethyl benzoic acid, 2-trifluoromethy-4-methybenzoic acid, 2-trifluoromethyl-4-methyl-benzoic acid, 4-methyl-2-trifluoromethyl-benzoic acid, 2-carboxy-5-methylbenzotrifluoride, 2-trifluoromethyl-p-toluic acid, 4-carboxy-3-trifluoromethyl toluene, 2-trifluoromethyl-4-methyl benzoic acid, 4-methyl-2-trifluoromethyl benzoicacid | |
4-methyl-2-(trifluoromethyl)benzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only