missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-(trimethylsilyl)phenyl Trifluoromethanesulfonate 98.0+%, TCI America™

$357.90 - $1267.56

Chemical Identifiers

CAS 262373-15-9
Molecular Formula C11H15F3O3SSi
Molecular Weight (g/mol) 312.378
InChI Key SKLVYEYSGSTOFD-UHFFFAOYSA-N
Synonym 4-Methyl-2-(trimethylsilyl)phenyl Triflate, Trifluoromethanesulfonic Acid 4-Methyl-2-(trimethylsilyl)phenyl Ester, 2-(Trimethylsilyl)-p-tolyl Trifluoromethanesulfonate
PubChem CID 11449792
IUPAC Name (4-methyl-2-trimethylsilylphenyl) trifluoromethanesulfonate
SMILES CC1=CC(=C(C=C1)OS(=O)(=O)C(F)(F)F)[Si](C)(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M18821G
View Documents
TCI America
M18821G
1 g
Each for $357.90
Only null left
 
M18825G
View Documents
TCI America
M18825G
5 g
Each for $1,267.56
Only null left
 
Description

Description

Specifications

Chemical Identifiers

262373-15-9
312.378
4-Methyl-2-(trimethylsilyl)phenyl Triflate, Trifluoromethanesulfonic Acid 4-Methyl-2-(trimethylsilyl)phenyl Ester, 2-(Trimethylsilyl)-p-tolyl Trifluoromethanesulfonate
(4-methyl-2-trimethylsilylphenyl) trifluoromethanesulfonate
C11H15F3O3SSi
SKLVYEYSGSTOFD-UHFFFAOYSA-N
11449792
CC1=CC(=C(C=C1)OS(=O)(=O)C(F)(F)F)[Si](C)(C)C

Specifications

262373-15-9
C11H15F3O3SSi
3265
SKLVYEYSGSTOFD-UHFFFAOYSA-N
(4-methyl-2-trimethylsilylphenyl) trifluoromethanesulfonate
11449792
≥98.0% (GC)
4-Methyl-2-(trimethylsilyl)phenyl Trifluoromethanesulfonate
Yellow
1 g
4-Methyl-2-(trimethylsilyl)phenyl Triflate, Trifluoromethanesulfonic Acid 4-Methyl-2-(trimethylsilyl)phenyl Ester, 2-(Trimethylsilyl)-p-tolyl Trifluoromethanesulfonate
CC1=CC(=C(C=C1)OS(=O)(=O)C(F)(F)F)[Si](C)(C)C
312.378
312.38
Liquid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'