Learn More
4-Methyl-3-nitrophenol, 98%, Thermo Scientific Chemicals
$118.18 - $294.96
Chemical Identifiers
CAS | 2042-14-0 |
---|---|
Molecular Formula | C7H7NO3 |
Molecular Weight (g/mol) | 153.137 |
MDL Number | MFCD00007244 |
InChI Key | BQEXDUKMTVYBRK-UHFFFAOYSA-N |
Synonym | 3-nitro-4-methylphenol, 3-nitro-p-cresol, 4-methyl-3-nitro-phenol, 4-hydroxy-2-nitrotoluene, 2-nitro-4-hydroxytoluene, phenol, 4-methyl-3-nitro, p-cresol, 3-nitro, unii-4b422hud6h, 4-methyl-5-nitrophenol |
PubChem CID | 16271 |
IUPAC Name | 4-methyl-3-nitrophenol |
SMILES | CC1=C(C=C(C=C1)O)[N+](=O)[O-] |
Description
It is used as a chemical, pharmaceutical and OLED intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2042-14-0 | |
153.137 | |
BQEXDUKMTVYBRK-UHFFFAOYSA-N | |
16271 | |
CC1=C(C=C(C=C1)O)[N+](=O)[O-] |
C7H7NO3 | |
MFCD00007244 | |
3-nitro-4-methylphenol, 3-nitro-p-cresol, 4-methyl-3-nitro-phenol, 4-hydroxy-2-nitrotoluene, 2-nitro-4-hydroxytoluene, phenol, 4-methyl-3-nitro, p-cresol, 3-nitro, unii-4b422hud6h, 4-methyl-5-nitrophenol | |
4-methyl-3-nitrophenol |
Specifications
2042-14-0 | |
Phenol-like | |
MFCD00007244 | |
UN2446 | |
Slightly soluble in water. | |
CC1=C(C=C(C=C1)O)[N+](=O)[O-] | |
153.137 | |
153.14 | |
Powder |
77°C to 81°C | |
C7H7NO3 | |
10 g | |
3-nitro-4-methylphenol, 3-nitro-p-cresol, 4-methyl-3-nitro-phenol, 4-hydroxy-2-nitrotoluene, 2-nitro-4-hydroxytoluene, phenol, 4-methyl-3-nitro, p-cresol, 3-nitro, unii-4b422hud6h, 4-methyl-5-nitrophenol | |
BQEXDUKMTVYBRK-UHFFFAOYSA-N | |
4-methyl-3-nitrophenol | |
16271 | |
98% | |
4-Methyl-3-nitrophenol |
Safety and Handling
GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITROCRESOLS, SOLID
EINECSNumber : 218-044-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only