missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Methyl-beta-styrylboronic acid pinacol ester, 98%, Thermo Scientific Chemicals
$179.89 - $477.05
Chemical Identifiers
CAS | 149777-84-4 |
---|---|
Molecular Formula | C15H21BO2 |
Molecular Weight (g/mol) | 244.141 |
MDL Number | MFCD03788749 |
InChI Key | HHBWKASJNTZJLB-ZHACJKMWSA-N |
Synonym | 4-methyl-beta-styrylboronic acid pinacol ester, e-4,4,5,5-tetramethyl-2-4-methylstyryl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-e-2-4-methylphenyl ethenyl-1,3,2-dioxaborolane, 4-methyl-b-styrylboronic acid pinacol ester, 4-methyl-, a-styrylboronic acid pinacol ester, 2-e-4-methylstyryl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, beta-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl 4-methylstyrene |
PubChem CID | 11042960 |
IUPAC Name | 4,4,5,5-tetramethyl-2-[(E)-2-(4-methylphenyl)ethenyl]-1,3,2-dioxaborolane |
SMILES | B1(OC(C(O1)(C)C)(C)C)C=CC2=CC=C(C=C2)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL19698MD
|
Thermo Scientific Chemicals
L19698MD |
250 mg |
Each for $179.89
|
|
|||||
AAL1969803
|
Thermo Scientific Chemicals
L1969803 |
1 g |
Each for $477.05
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
149777-84-4 | |
244.141 | |
HHBWKASJNTZJLB-ZHACJKMWSA-N | |
11042960 | |
B1(OC(C(O1)(C)C)(C)C)C=CC2=CC=C(C=C2)C |
C15H21BO2 | |
MFCD03788749 | |
4-methyl-beta-styrylboronic acid pinacol ester, e-4,4,5,5-tetramethyl-2-4-methylstyryl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-e-2-4-methylphenyl ethenyl-1,3,2-dioxaborolane, 4-methyl-b-styrylboronic acid pinacol ester, 4-methyl-, a-styrylboronic acid pinacol ester, 2-e-4-methylstyryl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, beta-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl 4-methylstyrene | |
4,4,5,5-tetramethyl-2-[(E)-2-(4-methylphenyl)ethenyl]-1,3,2-dioxaborolane |
Specifications
149777-84-4 | |
C15H21BO2 | |
250 mg | |
4-methyl-beta-styrylboronic acid pinacol ester, e-4,4,5,5-tetramethyl-2-4-methylstyryl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-e-2-4-methylphenyl ethenyl-1,3,2-dioxaborolane, 4-methyl-b-styrylboronic acid pinacol ester, 4-methyl-, a-styrylboronic acid pinacol ester, 2-e-4-methylstyryl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, beta-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl 4-methylstyrene | |
B1(OC(C(O1)(C)C)(C)C)C=CC2=CC=C(C=C2)C | |
244.141 | |
244.14 | |
4-Methyl-beta-styrylboronic acid pinacol ester |
56°C to 58°C | |
MFCD03788749 | |
7204616 | |
HHBWKASJNTZJLB-ZHACJKMWSA-N | |
4,4,5,5-tetramethyl-2-[(E)-2-(4-methylphenyl)ethenyl]-1,3,2-dioxaborolane | |
11042960 | |
98% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only