missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methyl-N-tosylbenzenesulfonamide 98.0+%, TCI America™
$76.43 - $229.29
Chemical Identifiers
| CAS | 3695-00-9 |
|---|---|
| Molecular Formula | C14H15NO4S2 |
| Molecular Weight (g/mol) | 325.397 |
| MDL Number | MFCD00014917 |
| InChI Key | LHWZLUXODWUHLZ-UHFFFAOYSA-N |
| Synonym | bis p-tolylsulfonyl amine, di-p-toluenesulfonamide, ditosylamine, di-p-toluenesulfonimide, bis 4-methylbenzenesulfon amine, benzenesulfonamide, 4-methyl-n-4-methylphenyl sulfonyl, 4-methyl-n-tosylbenzenesulfonamide, 4-methyl-n-4-methylphenyl sulfonyl benzenesulfonamide |
| PubChem CID | 77279 |
| IUPAC Name | 4-methyl-N-(4-methylphenyl)sulfonylbenzenesulfonamide |
| SMILES | CC1=CC=C(C=C1)S(=O)(=O)NS(=O)(=O)C2=CC=C(C=C2)C |
Chemical Identifiers
| 3695-00-9 | |
| 325.397 | |
| LHWZLUXODWUHLZ-UHFFFAOYSA-N | |
| 77279 | |
| CC1=CC=C(C=C1)S(=O)(=O)NS(=O)(=O)C2=CC=C(C=C2)C |
| C14H15NO4S2 | |
| MFCD00014917 | |
| bis p-tolylsulfonyl amine, di-p-toluenesulfonamide, ditosylamine, di-p-toluenesulfonimide, bis 4-methylbenzenesulfon amine, benzenesulfonamide, 4-methyl-n-4-methylphenyl sulfonyl, 4-methyl-n-tosylbenzenesulfonamide, 4-methyl-n-4-methylphenyl sulfonyl benzenesulfonamide | |
| 4-methyl-N-(4-methylphenyl)sulfonylbenzenesulfonamide |
Specifications
| 3695-00-9 | |
| White | |
| C14H15NO4S2 | |
| 1 g | |
| LHWZLUXODWUHLZ-UHFFFAOYSA-N | |
| 4-methyl-N-(4-methylphenyl)sulfonylbenzenesulfonamide | |
| 77279 | |
| ≥98.0% (HPLC) | |
| 4-Methyl-N-tosylbenzenesulfonamide |
| 173°C | |
| 265°C | |
| MFCD00014917 | |
| bis p-tolylsulfonyl amine, di-p-toluenesulfonamide, ditosylamine, di-p-toluenesulfonimide, bis 4-methylbenzenesulfon amine, benzenesulfonamide, 4-methyl-n-4-methylphenyl sulfonyl, 4-methyl-n-tosylbenzenesulfonamide, 4-methyl-n-4-methylphenyl sulfonyl benzenesulfonamide | |
| CC1=CC=C(C=C1)S(=O)(=O)NS(=O)(=O)C2=CC=C(C=C2)C | |
| 325.397 | |
| 325.40 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes