Learn More
4-(Methylamino)phenol sulfate, 99%, Thermo Scientific Chemicals
$50.10 - $467.81
Chemical Identifiers
CAS | 55-55-0 |
---|---|
Molecular Formula | C14H18N2O2·H2SO4 |
MDL Number | MFCD00013140 |
Synonym | Bis(4-hydroxy-N-methylanilinium) sulfate; Metol |
Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
AAA1623618
|
Thermo Scientific Chemicals
A1623618 |
50 g |
Each for $50.10
|
|
|||||
AAA1623630
|
Thermo Scientific Chemicals
A1623630 |
250 g |
Each for $155.53
|
|
|||||
AAA162360B
|
Thermo Scientific Chemicals
A162360B |
1000 g |
Each for $467.81
|
|
|||||
Description
4-(Methylamino)phenol sulfate, is a is a popular photographic developer used in black & white photography. It is also an excellent developing agent for most continuous tone developer applications, and it has been widely used in published developer formulas as well as commercial products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-(Methylamino)phenol sulfate, is a is a popular photographic developer used in black & white photography. It is also an excellent developing agent for most continuous tone developer applications, and it has been widely used in published developer formulas as well as commercial products.
Solubility
It is soluble in water (50 mg/mL).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from acids, acid chlorides, acid anhydrides, oxidizing agents.
Chemical Identifiers
55-55-0 | |
C14H18N2O2·H2SO4 | |
MFCD00013140 | |
Bis(4-hydroxy-N-methylanilinium) sulfate; Metol |
Specifications
55-55-0 | |
C14H18N2O2·H2SO4 | |
50 g | |
3919382 | |
14,6017 | |
It is soluble in water (50mg/mL). | |
99% |
∼260°C (decomposition) | |
MFCD00013140 | |
UN2811 | |
Air and light sensitive | |
Bis(4-hydroxy-N-methylanilinium) sulfate; Metol | |
344.38 | |
4-(Methylamino)phenol sulfate |
Safety and Handling
GHS H Statement
H373-H302-H317
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
May cause an allergic skin reaction.
P260-P264b-P270-P272-P280g-P301+P312-P302+P352-P314-P330-P333+P313-P363-P501c
H302-H317-H373-H500
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 200-237-1
RTECSNumber : SL8650000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only