missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Methylamino)pyridine, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1962603

 View more versions of this product

Catalog No. AAL1962603


Explore available promotions

Description

Description

4-(Methylamino)pyridine is employed as efficient nucleophilic catalyst during the preparation of ultra-high surface area emulsion templated porous polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Methylamino)pyridine is employed as efficient nucleophilic catalyst during the preparation of ultra-high surface area emulsion templated porous polymers.

Solubility
Soluble in water, acetone, ethanol, ether and benzene.

Notes
Keep the container tightly sealed. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

4-(Methylamino)pyridine
124°C to 128°C
MFCD00152625
108213
Soluble in water,acetone,ethanol,ether and benzene.
CNC1=CC=NC=C1
108.14
108.14
1121-58-0
C6H8N2
1 g
4-methylamino pyridine, 4-methylaminopyridine, n-methyl-4-pyridinamine, 4-pyridinamine, n-methyl, pyridin-4-yl methyl amine, 4-mamp, dmap on polystyrene, pubchem12915, pyridin-4yl-methylamine, pyridin-4-yl-methylamine
LSCYTCMNCWMCQE-UHFFFAOYSA-N
N-methylpyridin-4-amine
123098
99%

Chemical Identifiers

1121-58-0
108.14
LSCYTCMNCWMCQE-UHFFFAOYSA-N
123098
CNC1=CC=NC=C1
C6H8N2
MFCD00152625
4-methylamino pyridine, 4-methylaminopyridine, n-methyl-4-pyridinamine, 4-pyridinamine, n-methyl, pyridin-4-yl methyl amine, 4-mamp, dmap on polystyrene, pubchem12915, pyridin-4yl-methylamine, pyridin-4-yl-methylamine
N-methylpyridin-4-amine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only