missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methylanisole, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 164660010

 View more versions of this product

Catalog No. AC164660010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Methylanisole
98.5
-50°C
0.9600g/mL
53°C
98.5% min. (GC)
C8H10O
CH3C6H4OCH3
1 kg
0.96
Solubility in water: slightly soluble. Other solubilities: soluble in alchol
CC1=CC=C(C=C1)OC
122.17
122.17
Liquid
104-93-8
100.0
Colorless to Yellow
174°C
Authentic
Glass bottle
1.5100 to 1.513
MFCD00008413
06, 392
4-methylanisole, p-methylanisole, 4-methoxytoluene, p-methoxytoluene, benzene, 1-methoxy-4-methyl, p-cresyl methyl ether, p-cresol methyl ether, anisole, p-methyl, methyl p-tolyl ether, p-tolyl methyl ether
CHLICZRVGGXEOD-UHFFFAOYSA-N
1-methoxy-4-methylbenzene
7731
99%

Chemical Identifiers

104-93-8
122.17
CHLICZRVGGXEOD-UHFFFAOYSA-N
7731
CC1=CC=C(C=C1)OC
C8H10O
MFCD00008413
4-methylanisole, p-methylanisole, 4-methoxytoluene, p-methoxytoluene, benzene, 1-methoxy-4-methyl, p-cresyl methyl ether, p-cresol methyl ether, anisole, p-methyl, methyl p-tolyl ether, p-tolyl methyl ether
1-methoxy-4-methylbenzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Flammable liquid and vapor.
Causes skin irritation.
Suspected of damaging the unborn child.
Harmful to aquatic life with long lasting effects.

GHS P Statement
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing.

GHS Signal Word: Warning

EINECSNumber : 203-253-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only