Learn More
4-Methylanisole, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 164662500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Methylanisole | |
98.5 | |
-50°C | |
0.9600g/mL | |
53°C | |
98.5% min. (GC) | |
C8H10O | |
CH3C6H4OCH3 | |
250 g | |
0.96 | |
Solubility in water: slightly soluble. Other solubilities: soluble in alchol | |
CC1=CC=C(C=C1)OC | |
122.17 | |
122.17 | |
Liquid |
104-93-8 | |
100.0 | |
Colorless to Yellow | |
174°C | |
Authentic | |
Glass bottle | |
1.5100 to 1.513 | |
MFCD00008413 | |
06, 392 | |
4-methylanisole, p-methylanisole, 4-methoxytoluene, p-methoxytoluene, benzene, 1-methoxy-4-methyl, p-cresyl methyl ether, p-cresol methyl ether, anisole, p-methyl, methyl p-tolyl ether, p-tolyl methyl ether | |
CHLICZRVGGXEOD-UHFFFAOYSA-N | |
1-methoxy-4-methylbenzene | |
7731 | |
99% |
Chemical Identifiers
104-93-8 | |
122.17 | |
CHLICZRVGGXEOD-UHFFFAOYSA-N | |
7731 | |
CC1=CC=C(C=C1)OC |
C8H10O | |
MFCD00008413 | |
4-methylanisole, p-methylanisole, 4-methoxytoluene, p-methoxytoluene, benzene, 1-methoxy-4-methyl, p-cresyl methyl ether, p-cresol methyl ether, anisole, p-methyl, methyl p-tolyl ether, p-tolyl methyl ether | |
1-methoxy-4-methylbenzene |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Flammable liquid and vapor.
Causes skin irritation.
Suspected of damaging the unborn child.
Harmful to aquatic life with long lasting effects.
GHS P Statement
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing.
GHS Signal Word: Warning
EINECSNumber : 203-253-7
RUO – Research Use Only