Learn More
4-Methylbenzeneboronic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1334714
Description
4-Methylbenzeneboronic acid, is a commonly used chemical in Suzuki coupling reactions. It is also used as an intermediate in pharmaceutical industry
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Methylbenzeneboronic acid, is a commonly used chemical in Suzuki coupling reactions. It is also used as an intermediate in pharmaceutical industry
Solubility
It is soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications
4-Methylbenzeneboronic acid | |
246°C to 254°C | |
MFCD00236047 | |
2935970 | |
It is soluble in water. | |
COC1=CC=CC=C1B(O)O | |
151.96 | |
135.96 |
5720-05-8 | |
C7H9BO3 | |
25 g | |
2-methoxybenzeneboronic acid, 2-methoxyphenyl boronic acid, o-methoxyphenylboronic acid, 2-methoxyphenyl boranediol, 2-boronoanisole, 2-methoxypenylboronic acid, o-methoxyphenyl boronic acid, 2-methoxy phenylboronic acid, 2-methoxybenzene boronic acid | |
ROEQGIFOWRQYHD-UHFFFAOYSA-N | |
(2-methoxyphenyl)boronic acid | |
2733958 | |
98% |
Chemical Identifiers
5720-05-8 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
RTECSNumber : XS7400000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only