missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methylcatechol, 98%, Thermo Scientific Chemicals

$52.01 - $126.97

Chemical Identifiers

CAS 452-86-8
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.14
MDL Number MFCD00002205
InChI Key ZBCATMYQYDCTIZ-UHFFFAOYSA-N
Synonym 4-methylcatechol, 3,4-dihydroxytoluene, homocatechol, 4-methyl-1,2-benzenediol, p-methylcatechol, 4-methylpyrocatechol, homopyrocatechol, 1,2-dihydroxy-4-methylbenzene, p-methylpyrocatechol, toluene-3,4-diol
PubChem CID 9958
ChEBI CHEBI:17254
IUPAC Name 4-methylbenzene-1,2-diol
SMILES CC1=CC=C(O)C(O)=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC126510250
SDS View Documents
Thermo Scientific Chemicals
126510250
25 g Glass bottle
Each for $52.01
 
AC126511000
SDS View Documents
Thermo Scientific Chemicals
126511000
100 g Glass bottle
Each for $126.97
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

452-86-8
124.14
ZBCATMYQYDCTIZ-UHFFFAOYSA-N
9958
4-methylbenzene-1,2-diol
C7H8O2
MFCD00002205
4-methylcatechol, 3,4-dihydroxytoluene, homocatechol, 4-methyl-1,2-benzenediol, p-methylcatechol, 4-methylpyrocatechol, homopyrocatechol, 1,2-dihydroxy-4-methylbenzene, p-methylpyrocatechol, toluene-3,4-diol
CHEBI:17254
CC1=CC=C(O)C(O)=C1

Specifications

452-86-8
100.0
Brown to White
140°C
97.5% min. (GC)
C7H8O2
MFCD00002205
06, 878
Solubility in water: soluble in water. Other solubilities: miscible with acetone,alcohol,ether and toluene,moderately soluble in petroleum ether,slightly soluble in carbon tetrachloride
CC1=CC=C(O)C(O)=C1
124.14
CHEBI:17254
98%
4-Methylcatechol
97.5
62°C to 69°C
251°C
Authentic
Glass bottle
CH3C6H3(OH)2
25 g
4-methylcatechol, 3,4-dihydroxytoluene, homocatechol, 4-methyl-1,2-benzenediol, p-methylcatechol, 4-methylpyrocatechol, homopyrocatechol, 1,2-dihydroxy-4-methylbenzene, p-methylpyrocatechol, toluene-3,4-diol
ZBCATMYQYDCTIZ-UHFFFAOYSA-N
4-methylbenzene-1,2-diol
9958
124.14
Chunks and/or Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

GHS Signal Word: Warning

EINECSNumber : 207-214-5

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only