missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methylcyclohexanone, 98%, Thermo Scientific Chemicals

$83.85 - $83.85

Chemical Identifiers

CAS 589-92-4
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.17
MDL Number MFCD00001643
InChI Key VGVHNLRUAMRIEW-UHFFFAOYSA-N
Synonym 4-methylcyclohexanone, cyclohexanone, 4-methyl, 4-methyl-1-cyclohexanone, methyl-4 cyclohexanone-1, p-methyl cyclohexanone, methycyclohexanone, metylocykloheksanon, unii-0l1r78k79s, 4-methyl cyclohexanone, methyl-4 cyclohexanone-1 french
PubChem CID 11525
IUPAC Name 4-methylcyclohexan-1-one
SMILES CC1CCC(=O)CC1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC174260250
SDS View Documents
Thermo Scientific Chemicals
174260250
25 g N/A
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

589-92-4
112.17
VGVHNLRUAMRIEW-UHFFFAOYSA-N
11525
CC1CCC(=O)CC1
C7H12O
MFCD00001643
4-methylcyclohexanone, cyclohexanone, 4-methyl, 4-methyl-1-cyclohexanone, methyl-4 cyclohexanone-1, p-methyl cyclohexanone, methycyclohexanone, metylocykloheksanon, unii-0l1r78k79s, 4-methyl cyclohexanone, methyl-4 cyclohexanone-1 french
4-methylcyclohexan-1-one

Specifications

589-92-4
100.0
Colorless to Yellow
169°C to 171°C
Authentic
Glass bottle
1.4445 to 1.4465
MFCD00001643
07,18
4-methylcyclohexanone, cyclohexanone, 4-methyl, 4-methyl-1-cyclohexanone, methyl-4 cyclohexanone-1, p-methyl cyclohexanone, methycyclohexanone, metylocykloheksanon, unii-0l1r78k79s, 4-methyl cyclohexanone, methyl-4 cyclohexanone-1 french
CC1CCC(=O)CC1
112.17
112.17
Liquid
97.5
-41°C
0.9140g/mL
40°C
98%
C7H12O
CH3C6H9(=O)
25 g
0.914
VGVHNLRUAMRIEW-UHFFFAOYSA-N
4-methylcyclohexan-1-one
11525
98%
4-Methylcyclohexanone, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 209-665-3

RTECSNumber : GW1750200

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only