Learn More
4-Methylcyclohexanone, 98+%, Thermo Scientific Chemicals
$209.60 - $209.60
Chemical Identifiers
CAS | 589-92-4 |
---|---|
Molecular Formula | C7H12O |
Molecular Weight (g/mol) | 112.17 |
MDL Number | MFCD00001643 |
InChI Key | VGVHNLRUAMRIEW-UHFFFAOYSA-N |
Synonym | 4-methylcyclohexanone, cyclohexanone, 4-methyl, 4-methyl-1-cyclohexanone, methyl-4 cyclohexanone-1, p-methyl cyclohexanone, methycyclohexanone, metylocykloheksanon, unii-0l1r78k79s, 4-methyl cyclohexanone, methyl-4 cyclohexanone-1 french |
PubChem CID | 11525 |
IUPAC Name | 4-methylcyclohexan-1-one |
SMILES | CC1CCC(=O)CC1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1318622
|
Thermo Scientific Chemicals
A1318622 |
100 g |
Each for $209.60
|
|
|||||
Description
4-Methylcyclohexanone was used in the synthesis of the pinguisane-type sesquiterpenoid acutifolone A, in racemic form. It is used as intermediates of CCNU and it can be also be used as a solvent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Methylcyclohexanone was used in the synthesis of the pinguisane-type sesquiterpenoid acutifolone A, in racemic form. It is used as intermediates of CCNU and it can be also be used as a solvent.
Solubility
Not miscible in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
589-92-4 | |
112.17 | |
VGVHNLRUAMRIEW-UHFFFAOYSA-N | |
11525 | |
CC1CCC(=O)CC1 |
C7H12O | |
MFCD00001643 | |
4-methylcyclohexanone, cyclohexanone, 4-methyl, 4-methyl-1-cyclohexanone, methyl-4 cyclohexanone-1, p-methyl cyclohexanone, methycyclohexanone, metylocykloheksanon, unii-0l1r78k79s, 4-methyl cyclohexanone, methyl-4 cyclohexanone-1 french | |
4-methylcyclohexan-1-one |
Specifications
589-92-4 | |
0.914 | |
48°C (118°F) | |
1.445 | |
100 g | |
506746 | |
Not miscible in water. | |
CC1CCC(=O)CC1 | |
112.17 | |
112.17 | |
4-Methylcyclohexanone |
-41°C | |
169°C to 171°C | |
C7H12O | |
MFCD00001643 | |
UN2297 | |
4-methylcyclohexanone, cyclohexanone, 4-methyl, 4-methyl-1-cyclohexanone, methyl-4 cyclohexanone-1, p-methyl cyclohexanone, methycyclohexanone, metylocykloheksanon, unii-0l1r78k79s, 4-methyl cyclohexanone, methyl-4 cyclohexanone-1 french | |
VGVHNLRUAMRIEW-UHFFFAOYSA-N | |
4-methylcyclohexan-1-one | |
11525 | |
≥98% |
Safety and Handling
GHS H Statement
H301-H226
Toxic if swallowed.
Flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P330-P363-P370+P378q-P501c
H226-H302+H312+H332
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: METHYLCYCLOHEXANONE
EINECSNumber : 209-665-3
RTECSNumber : GW1750200
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only