missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methylimidazole, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 190332500

Catalog No. AC190332500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

4-Methylimidazole
822-36-6
100.0
157°C
97.5% min. (GC)
C4H6N2
250g
4-methylimidazole, 4-methyl-1h-imidazole, 4 5-methylimidazole, 1h-imidazole, 4-methyl, 4-me-i, 5-methylimidazole, imidazole, 4-methyl, 4 or 5-methylimidazole, 4-mei, 4-methylimidazol
XLSZMDLNRCVEIJ-UHFFFAOYSA-N
5-methyl-1H-imidazole
13195
82.11
98%
97.5
263.0°C
Authentic
Glass bottle
MFCD00005201
23, 69
Solubility in water: soluble. Other solubilities: soluble in almost all common organic solvents
CC1=CN=CN1
82.11
CHEBI:40035
98%
Safety and Handling

Safety and Handling

GHS H Statement
Toxic in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.

GHS Signal Word: Danger

EINECSNumber : 212-497-3

RTECSNumber : NI7350000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only